Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 100908305-100908574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1A3
Entrez Gene ID
220
Gene Name
aldehyde dehydrogenase 1 family member A3
Gene Aliases
ALDH1A6, ALDH6, MCOP8, RALDH3
UniGene
Hs.459538
Gene Chromosome Location
Chr.15: 100879692-100916626 on Build GRCh38
Transcripts
NM_000693.3 NM_001293815.1 NR_135827.1

Gene Symbol
LOC101927751
Entrez Gene ID
101927751
Gene Name
uncharacterized LOC101927751
Gene Aliases
-
UniGene
Hs.659224
Gene Chromosome Location
Chr.15: 100892343-100919283 on Build GRCh38
Transcripts
NM_000693.3 NM_001293815.1 NR_135827.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5580188 c.1433G>T p.G478V Chr.15: 100908449-100908449 on Build GRCh38
4661003 c.1407C>T p.N469N Chr.15: 100908423-100908423 on Build GRCh38
1375684 c.1408G>A p.A470T Chr.15: 100908424-100908424 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757881921 Chr.15: 100908500-100908500 on Build GRCh38
rs145132174 Chr.15: 100908533-100908533 on Build GRCh38
rs879424694 Chr.15: 100908334-100908334 on Build GRCh38
rs189759261 Chr.15: 100908512-100908512 on Build GRCh38
rs745865485 Chr.15: 100908514-100908514 on Build GRCh38
rs550784570 Chr.15: 100908404-100908404 on Build GRCh38
rs372547390 Chr.15: 100908390-100908390 on Build GRCh38
rs147752643 Chr.15: 100908424-100908424 on Build GRCh38
rs760973105 Chr.15: 100908451-100908451 on Build GRCh38
rs947226082 Chr.15: 100908449-100908449 on Build GRCh38
rs371573786 Chr.15: 100908355-100908355 on Build GRCh38
rs199824298 Chr.15: 100908369-100908369 on Build GRCh38
rs200617229 Chr.15: 100908474-100908474 on Build GRCh38
rs772146905 Chr.15: 100908524-100908524 on Build GRCh38
rs200510714 Chr.15: 100908396-100908396 on Build GRCh38
rs201444877 Chr.15: 100908481-100908481 on Build GRCh38
rs745736697 Chr.15: 100908392-100908392 on Build GRCh38
rs747191192 Chr.15: 100908531-100908531 on Build GRCh38
rs4646686 Chr.15: 100908366-100908366 on Build GRCh38
rs777111865 Chr.15: 100908534-100908534 on Build GRCh38
rs749120702 Chr.15: 100908430-100908431 on Build GRCh38
rs768700601 Chr.15: 100908335-100908336 on Build GRCh38
rs773178840 Chr.15: 100908361-100908361 on Build GRCh38
rs377069366 Chr.15: 100908506-100908506 on Build GRCh38
rs773322840 Chr.15: 100908528-100908528 on Build GRCh38
rs766710815 Chr.15: 100908459-100908459 on Build GRCh38
rs376447003 Chr.15: 100908519-100908519 on Build GRCh38
rs145659258 Chr.15: 100908423-100908423 on Build GRCh38
rs920204189 Chr.15: 100908345-100908345 on Build GRCh38
rs752220935 Chr.15: 100908497-100908497 on Build GRCh38
rs756023264 Chr.15: 100908517-100908517 on Build GRCh38
rs915715572 Chr.15: 100908436-100908436 on Build GRCh38
rs777285496 Chr.15: 100908471-100908471 on Build GRCh38
rs777659338 Chr.15: 100908363-100908363 on Build GRCh38
rs898308107 Chr.15: 100908442-100908442 on Build GRCh38
rs773448078 Chr.15: 100908435-100908435 on Build GRCh38
rs772154609 Chr.15: 100908427-100908427 on Build GRCh38
rs757961323 Chr.15: 100908362-100908362 on Build GRCh38
rs370199725 Chr.15: 100908511-100908511 on Build GRCh38

View Full Product Details