Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33223778-33224021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFA3
Entrez Gene ID
128861
Gene Name
BPI fold containing family A member 3
Gene Aliases
C20orf71, SPLUNC3
UniGene
Hs.360989
Gene Chromosome Location
Chr.20: 33217310-33227806 on Build GRCh38
Transcripts
NM_001042439.2 NM_178466.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1411180 c.251G>T p.R84M Chr.20: 33223934-33223934 on Build GRCh38
397131 c.128-1G>A p.? Chr.20: 33223810-33223810 on Build GRCh38
7136056 c.144C>A p.L48L Chr.20: 33223827-33223827 on Build GRCh38
3423588 c.129T>G p.I43M Chr.20: 33223812-33223812 on Build GRCh38
7117739 c.230T>C p.V77A Chr.20: 33223913-33223913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887887978 Chr.20: 33223847-33223847 on Build GRCh38
rs758368178 Chr.20: 33223839-33223839 on Build GRCh38
rs574453101 Chr.20: 33223909-33223909 on Build GRCh38
rs199537127 Chr.20: 33223972-33223972 on Build GRCh38
rs149822522 Chr.20: 33223871-33223871 on Build GRCh38
rs1040575897 Chr.20: 33223826-33223826 on Build GRCh38
rs374504938 Chr.20: 33223935-33223935 on Build GRCh38
rs112370343 Chr.20: 33223928-33223928 on Build GRCh38
rs767047370 Chr.20: 33223880-33223880 on Build GRCh38
rs537299136 Chr.20: 33223968-33223968 on Build GRCh38
rs532125562 Chr.20: 33223840-33223840 on Build GRCh38
rs753109478 Chr.20: 33223982-33223982 on Build GRCh38
rs144223805 Chr.20: 33223943-33223943 on Build GRCh38
rs150331436 Chr.20: 33223849-33223849 on Build GRCh38
rs757165126 Chr.20: 33223921-33223921 on Build GRCh38
rs751696718 Chr.20: 33223916-33223916 on Build GRCh38
rs201261464 Chr.20: 33223862-33223862 on Build GRCh38
rs373502234 Chr.20: 33223799-33223799 on Build GRCh38
rs764349800 Chr.20: 33223990-33223990 on Build GRCh38
rs758659643 Chr.20: 33223903-33223903 on Build GRCh38
rs200445146 Chr.20: 33223887-33223887 on Build GRCh38
rs767997234 Chr.20: 33223872-33223872 on Build GRCh38
rs866768394 Chr.20: 33223834-33223834 on Build GRCh38
rs187833570 Chr.20: 33223971-33223971 on Build GRCh38
rs779915382 Chr.20: 33223930-33223930 on Build GRCh38
rs767892601 Chr.20: 33223824-33223824 on Build GRCh38
rs756677564 Chr.20: 33223900-33223900 on Build GRCh38
rs765865186 Chr.20: 33223835-33223835 on Build GRCh38
rs367718982 Chr.20: 33223948-33223948 on Build GRCh38
rs113151188 Chr.20: 33223929-33223929 on Build GRCh38
rs781149278 Chr.20: 33223927-33223927 on Build GRCh38
rs1006604225 Chr.20: 33223888-33223888 on Build GRCh38
rs774969132 Chr.20: 33223869-33223869 on Build GRCh38
rs748829090 Chr.20: 33223931-33223931 on Build GRCh38
rs779049750 Chr.20: 33223823-33223823 on Build GRCh38
rs139890063 Chr.20: 33223820-33223820 on Build GRCh38
rs372091970 Chr.20: 33223970-33223970 on Build GRCh38
rs373749585 Chr.20: 33223992-33223992 on Build GRCh38
rs761324963 Chr.20: 33223807-33223807 on Build GRCh38
rs771745365 Chr.20: 33223848-33223848 on Build GRCh38
rs745713946 Chr.20: 33223861-33223861 on Build GRCh38
rs190813318 Chr.20: 33223810-33223810 on Build GRCh38
rs754230263 Chr.20: 33223883-33223883 on Build GRCh38
rs562818353 Chr.20: 33223828-33223828 on Build GRCh38
rs996049237 Chr.20: 33223829-33223829 on Build GRCh38
rs750803835 Chr.20: 33223850-33223850 on Build GRCh38
rs761454800 Chr.20: 33223942-33223942 on Build GRCh38
rs770921952 Chr.20: 33223846-33223846 on Build GRCh38
rs551941343 Chr.20: 33223868-33223868 on Build GRCh38
rs1043428271 Chr.20: 33223803-33223803 on Build GRCh38
rs774006719 Chr.20: 33223879-33223879 on Build GRCh38
rs148580329 Chr.20: 33223941-33223941 on Build GRCh38
rs565577075 Chr.20: 33223893-33223893 on Build GRCh38

View Full Product Details