Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 25377363-25377626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC4
Entrez Gene ID
29945
Gene Name
anaphase promoting complex subunit 4
Gene Aliases
APC4
UniGene
Hs.152173
Gene Chromosome Location
Chr.4: 25377226-25418498 on Build GRCh38
Transcripts
NM_001286756.1 NM_013367.2 XM_011513838.1 XM_011513839.1 XM_017008138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3603080 c.90C>T p.P30P Chr.4: 25377517-25377517 on Build GRCh38
4517272 c.29_30CC>AT p.S10Y Chr.4: 25377456-25377457 on Build GRCh38
288052 c.81C>T p.V27V Chr.4: 25377508-25377508 on Build GRCh38
6203788 c.69_70delTA p.I24fs*38 Chr.4: 25377496-25377497 on Build GRCh38
5889984 c.129+7G>A p.? Chr.4: 25377563-25377563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563308193 Chr.4: 25377540-25377540 on Build GRCh38
rs35359297 Chr.4: 25377441-25377442 on Build GRCh38
rs774607747 Chr.4: 25377402-25377403 on Build GRCh38
rs374595036 Chr.4: 25377488-25377488 on Build GRCh38
rs543616759 Chr.4: 25377456-25377456 on Build GRCh38
rs373808140 Chr.4: 25377406-25377406 on Build GRCh38
rs776665542 Chr.4: 25377603-25377603 on Build GRCh38
rs759844168 Chr.4: 25377570-25377570 on Build GRCh38
rs953985592 Chr.4: 25377590-25377590 on Build GRCh38
rs772072500 Chr.4: 25377449-25377449 on Build GRCh38
rs767625635 Chr.4: 25377501-25377501 on Build GRCh38
rs771088242 Chr.4: 25377599-25377599 on Build GRCh38
rs6830160 Chr.4: 25377604-25377604 on Build GRCh38
rs988784191 Chr.4: 25377386-25377386 on Build GRCh38
rs747026168 Chr.4: 25377596-25377596 on Build GRCh38
rs748595415 Chr.4: 25377575-25377575 on Build GRCh38
rs762869890 Chr.4: 25377383-25377383 on Build GRCh38
rs768022403 Chr.4: 25377574-25377623 on Build GRCh38
rs796663160 Chr.4: 25377395-25377395 on Build GRCh38
rs755691767 Chr.4: 25377538-25377538 on Build GRCh38
rs748685739 Chr.4: 25377435-25377435 on Build GRCh38
rs761858918 Chr.4: 25377498-25377498 on Build GRCh38
rs750043555 Chr.4: 25377403-25377403 on Build GRCh38
rs773292926 Chr.4: 25377454-25377454 on Build GRCh38
rs778317686 Chr.4: 25377580-25377580 on Build GRCh38
rs749586422 Chr.4: 25377411-25377411 on Build GRCh38
rs771587730 Chr.4: 25377543-25377543 on Build GRCh38
rs577496521 Chr.4: 25377420-25377420 on Build GRCh38
rs540624104 Chr.4: 25377397-25377397 on Build GRCh38
rs747148002 Chr.4: 25377455-25377455 on Build GRCh38
rs370977952 Chr.4: 25377387-25377387 on Build GRCh38
rs755259219 Chr.4: 25377555-25377555 on Build GRCh38
rs756605519 Chr.4: 25377609-25377609 on Build GRCh38
rs370535597 Chr.4: 25377576-25377576 on Build GRCh38
rs776949076 Chr.4: 25377475-25377475 on Build GRCh38
rs947104282 Chr.4: 25377404-25377404 on Build GRCh38
rs1027340582 Chr.4: 25377482-25377482 on Build GRCh38
rs780857667 Chr.4: 25377399-25377399 on Build GRCh38
rs199602455 Chr.4: 25377410-25377410 on Build GRCh38
rs751592232 Chr.4: 25377391-25377391 on Build GRCh38
rs762628911 Chr.4: 25377484-25377484 on Build GRCh38
rs749952036 Chr.4: 25377529-25377529 on Build GRCh38
rs557655484 Chr.4: 25377415-25377415 on Build GRCh38
rs774367060 Chr.4: 25377491-25377491 on Build GRCh38
rs779252233 Chr.4: 25377570-25377570 on Build GRCh38
rs1051893863 Chr.4: 25377494-25377494 on Build GRCh38
rs763969481 Chr.4: 25377487-25377487 on Build GRCh38
rs529390900 Chr.4: 25377545-25377545 on Build GRCh38
rs753134191 Chr.4: 25377584-25377585 on Build GRCh38
rs752419950 Chr.4: 25377381-25377381 on Build GRCh38

View Full Product Details