Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7075289-7075520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC10A
Entrez Gene ID
10462
Gene Name
C-type lectin domain family 10 member A
Gene Aliases
CD301, CLECSF13, CLECSF14, HML, HML2, MGL
UniGene
Hs.54403
Gene Chromosome Location
Chr.17: 7074537-7080635 on Build GRCh38
Transcripts
NM_006344.2 NM_182906.2 XM_005256412.1 XM_011523613.1 XM_011523614.2 XM_011523615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5913862 c.727G>A p.D243N Chr.17: 7075415-7075415 on Build GRCh38
378683 c.776G>T p.G259V Chr.17: 7075366-7075366 on Build GRCh38
4672707 c.770C>T p.A257V Chr.17: 7075372-7075372 on Build GRCh38
6630074 c.782+6T>C p.? Chr.17: 7075354-7075354 on Build GRCh38
1709693 c.763G>A p.D255N Chr.17: 7075379-7075379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778981235 Chr.17: 7075326-7075326 on Build GRCh38
rs377248841 Chr.17: 7075317-7075317 on Build GRCh38
rs958419245 Chr.17: 7075411-7075411 on Build GRCh38
rs772347436 Chr.17: 7075463-7075474 on Build GRCh38
rs572028091 Chr.17: 7075491-7075491 on Build GRCh38
rs773718598 Chr.17: 7075389-7075389 on Build GRCh38
rs978312083 Chr.17: 7075482-7075482 on Build GRCh38
rs957227750 Chr.17: 7075425-7075425 on Build GRCh38
rs751929611 Chr.17: 7075382-7075382 on Build GRCh38
rs759724775 Chr.17: 7075434-7075434 on Build GRCh38
rs140041915 Chr.17: 7075374-7075374 on Build GRCh38
rs749816425 Chr.17: 7075500-7075500 on Build GRCh38
rs767516359 Chr.17: 7075319-7075319 on Build GRCh38
rs190240638 Chr.17: 7075469-7075469 on Build GRCh38
rs774649349 Chr.17: 7075427-7075427 on Build GRCh38
rs370937777 Chr.17: 7075478-7075478 on Build GRCh38
rs150484759 Chr.17: 7075448-7075448 on Build GRCh38
rs752798672 Chr.17: 7075323-7075323 on Build GRCh38
rs62619837 Chr.17: 7075358-7075358 on Build GRCh38
rs989509836 Chr.17: 7075452-7075452 on Build GRCh38
rs766765887 Chr.17: 7075368-7075368 on Build GRCh38
rs752777715 Chr.17: 7075440-7075440 on Build GRCh38
rs747696083 Chr.17: 7075490-7075490 on Build GRCh38
rs770259937 Chr.17: 7075372-7075372 on Build GRCh38
rs767775644 Chr.17: 7075439-7075439 on Build GRCh38
rs746792236 Chr.17: 7075473-7075473 on Build GRCh38
rs758273609 Chr.17: 7075329-7075329 on Build GRCh38
rs939574132 Chr.17: 7075493-7075493 on Build GRCh38
rs777223465 Chr.17: 7075498-7075498 on Build GRCh38
rs779957846 Chr.17: 7075350-7075350 on Build GRCh38
rs763383216 Chr.17: 7075417-7075417 on Build GRCh38
rs140313272 Chr.17: 7075470-7075470 on Build GRCh38
rs368116399 Chr.17: 7075324-7075324 on Build GRCh38
rs766758789 Chr.17: 7075419-7075419 on Build GRCh38
rs1034493844 Chr.17: 7075405-7075405 on Build GRCh38
rs17853871 Chr.17: 7075446-7075446 on Build GRCh38
rs898011432 Chr.17: 7075349-7075349 on Build GRCh38
rs556438794 Chr.17: 7075327-7075327 on Build GRCh38
rs768336086 Chr.17: 7075367-7075367 on Build GRCh38
rs1007269983 Chr.17: 7075375-7075375 on Build GRCh38
rs578067885 Chr.17: 7075355-7075355 on Build GRCh38
rs747685144 Chr.17: 7075371-7075371 on Build GRCh38
rs924418589 Chr.17: 7075423-7075423 on Build GRCh38
rs977381058 Chr.17: 7075413-7075413 on Build GRCh38
rs368562852 Chr.17: 7075471-7075471 on Build GRCh38

View Full Product Details