Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160115737-160115998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A2
Entrez Gene ID
477
Gene Name
ATPase Na+/K+ transporting subunit alpha 2
Gene Aliases
FHM2, MHP2
UniGene
Hs.34114
Gene Chromosome Location
Chr.1: 160115730-160143591 on Build GRCh38
Transcripts
NM_000702.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
222458 c.5G>A p.G2D Chr.1: 160115866-160115866 on Build GRCh38
3385332 c.8G>A p.R3H Chr.1: 160115869-160115869 on Build GRCh38
6214280 c.11G>T p.G4V Chr.1: 160115872-160115872 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774433663 Chr.1: 160115821-160115821 on Build GRCh38
rs373105548 Chr.1: 160115900-160115900 on Build GRCh38
rs759542448 Chr.1: 160115832-160115832 on Build GRCh38
rs978766277 Chr.1: 160115809-160115809 on Build GRCh38
rs41265761 Chr.1: 160115814-160115814 on Build GRCh38
rs888389853 Chr.1: 160115879-160115879 on Build GRCh38
rs989024254 Chr.1: 160115771-160115771 on Build GRCh38
rs781687346 Chr.1: 160115869-160115869 on Build GRCh38
rs186666025 Chr.1: 160115918-160115918 on Build GRCh38
rs780391321 Chr.1: 160115850-160115850 on Build GRCh38
rs182380403 Chr.1: 160115833-160115833 on Build GRCh38
rs374133858 Chr.1: 160115963-160115963 on Build GRCh38
rs937616856 Chr.1: 160115798-160115798 on Build GRCh38
rs753471059 Chr.1: 160115844-160115844 on Build GRCh38
rs772637490 Chr.1: 160115920-160115920 on Build GRCh38
rs377546471 Chr.1: 160115902-160115902 on Build GRCh38
rs755450946 Chr.1: 160115868-160115868 on Build GRCh38
rs192111085 Chr.1: 160115831-160115831 on Build GRCh38
rs369670270 Chr.1: 160115897-160115897 on Build GRCh38
rs146635406 Chr.1: 160115964-160115964 on Build GRCh38
rs376784470 Chr.1: 160115885-160115885 on Build GRCh38
rs546037926 Chr.1: 160115968-160115968 on Build GRCh38
rs564645266 Chr.1: 160115972-160115972 on Build GRCh38
rs756960901 Chr.1: 160115848-160115848 on Build GRCh38
rs769847812 Chr.1: 160115890-160115890 on Build GRCh38
rs371023222 Chr.1: 160115842-160115842 on Build GRCh38
rs377605497 Chr.1: 160115813-160115813 on Build GRCh38
rs775459753 Chr.1: 160115913-160115913 on Build GRCh38
rs535758394 Chr.1: 160115930-160115930 on Build GRCh38
rs1005907824 Chr.1: 160115896-160115896 on Build GRCh38
rs569640199 Chr.1: 160115792-160115792 on Build GRCh38
rs751958298 Chr.1: 160115860-160115860 on Build GRCh38
rs1018043439 Chr.1: 160115912-160115913 on Build GRCh38
rs772016755 Chr.1: 160115912-160115912 on Build GRCh38
rs774258321 Chr.1: 160115909-160115909 on Build GRCh38
rs759522814 Chr.1: 160115916-160115916 on Build GRCh38
rs981682686 Chr.1: 160115778-160115778 on Build GRCh38
rs913496641 Chr.1: 160115780-160115780 on Build GRCh38
rs755602176 Chr.1: 160115838-160115838 on Build GRCh38
rs372906753 Chr.1: 160115953-160115953 on Build GRCh38
rs748630987 Chr.1: 160115886-160115886 on Build GRCh38
rs759452586 Chr.1: 160115910-160115910 on Build GRCh38
rs532142136 Chr.1: 160115881-160115881 on Build GRCh38

View Full Product Details