Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35026299-35026568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGXT2
Entrez Gene ID
64902
Gene Name
alanine--glyoxylate aminotransferase 2
Gene Aliases
AGT2, DAIBAT
UniGene
Hs.34494
Gene Chromosome Location
Chr.5: 34998101-35048188 on Build GRCh38
Transcripts
NM_001306173.1 NM_031900.3 XM_005248337.2 XM_005248338.2 XM_017009748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5746398 c.814A>G p.T272A Chr.5: 35026466-35026466 on Build GRCh38
3994292 c.772T>G p.C258G Chr.5: 35026508-35026508 on Build GRCh38
5975588 c.780A>G p.Q260Q Chr.5: 35026500-35026500 on Build GRCh38
3854859 c.813T>C p.D271D Chr.5: 35026467-35026467 on Build GRCh38
391281 c.838_847del10 p.S280fs*10 Chr.5: 35026433-35026442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778352603 Chr.5: 35026435-35026436 on Build GRCh38
rs775148379 Chr.5: 35026431-35026431 on Build GRCh38
rs752043415 Chr.5: 35026458-35026458 on Build GRCh38
rs73084607 Chr.5: 35026336-35026336 on Build GRCh38
rs759333159 Chr.5: 35026405-35026405 on Build GRCh38
rs756531820 Chr.5: 35026396-35026396 on Build GRCh38
rs930170136 Chr.5: 35026417-35026417 on Build GRCh38
rs758998542 Chr.5: 35026452-35026452 on Build GRCh38
rs137970182 Chr.5: 35026364-35026364 on Build GRCh38
rs755422899 Chr.5: 35026464-35026464 on Build GRCh38
rs200638973 Chr.5: 35026542-35026542 on Build GRCh38
rs144708069 Chr.5: 35026500-35026500 on Build GRCh38
rs185340432 Chr.5: 35026471-35026471 on Build GRCh38
rs766805931 Chr.5: 35026456-35026456 on Build GRCh38
rs895855552 Chr.5: 35026495-35026495 on Build GRCh38
rs756918425 Chr.5: 35026434-35026435 on Build GRCh38
rs985539770 Chr.5: 35026361-35026361 on Build GRCh38
rs561547784 Chr.5: 35026486-35026486 on Build GRCh38
rs749341798 Chr.5: 35026503-35026503 on Build GRCh38
rs149356098 Chr.5: 35026445-35026445 on Build GRCh38
rs748513688 Chr.5: 35026378-35026378 on Build GRCh38
rs777514053 Chr.5: 35026384-35026384 on Build GRCh38
rs774306394 Chr.5: 35026514-35026514 on Build GRCh38
rs745348522 Chr.5: 35026525-35026526 on Build GRCh38
rs572826520 Chr.5: 35026465-35026465 on Build GRCh38
rs748833390 Chr.5: 35026413-35026427 on Build GRCh38
rs778109101 Chr.5: 35026504-35026504 on Build GRCh38
rs771906704 Chr.5: 35026535-35026535 on Build GRCh38
rs1049297283 Chr.5: 35026447-35026447 on Build GRCh38
rs773142699 Chr.5: 35026381-35026381 on Build GRCh38
rs565080389 Chr.5: 35026374-35026374 on Build GRCh38
rs1032116566 Chr.5: 35026324-35026324 on Build GRCh38
rs375279525 Chr.5: 35026521-35026521 on Build GRCh38
rs533299831 Chr.5: 35026425-35026425 on Build GRCh38
rs911901736 Chr.5: 35026341-35026341 on Build GRCh38
rs760447666 Chr.5: 35026545-35026545 on Build GRCh38
rs192883427 Chr.5: 35026399-35026399 on Build GRCh38
rs760225755 Chr.5: 35026439-35026439 on Build GRCh38

View Full Product Details