Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122981582-122981855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL6IP4
Entrez Gene ID
51329
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 4
Gene Aliases
SFRS20, SR-25, SRp25, SRrp37
UniGene
Hs.103561
Gene Chromosome Location
Chr.12: 122980060-122982913 on Build GRCh38
Transcripts
NM_001002251.2 NM_001002252.2 NM_001278378.1 NM_001278379.1 NM_001278380.1 NM_016638.3 NM_018694.3 NR_103512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5644804 c.548C>T p.S183F Chr.12: 122981646-122981646 on Build GRCh38
5374483 c.625G>A p.G209R Chr.12: 122981723-122981723 on Build GRCh38
3954318 c.700A>T p.R234W Chr.12: 122981741-122981741 on Build GRCh38
5970568 c.654G>A p.K218K Chr.12: 122981752-122981752 on Build GRCh38
3720872 c.641C>T p.S214L Chr.12: 122981682-122981682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779795298 Chr.12: 122981673-122981673 on Build GRCh38
rs146762957 Chr.12: 122981764-122981764 on Build GRCh38
rs548529193 Chr.12: 122981691-122981691 on Build GRCh38
rs746399440 Chr.12: 122981783-122981783 on Build GRCh38
rs777430007 Chr.12: 122981720-122981721 on Build GRCh38
rs375563649 Chr.12: 122981713-122981713 on Build GRCh38
rs763691508 Chr.12: 122981647-122981647 on Build GRCh38
rs766112918 Chr.12: 122981824-122981824 on Build GRCh38
rs952323363 Chr.12: 122981688-122981688 on Build GRCh38
rs759474425 Chr.12: 122981834-122981834 on Build GRCh38
rs117153243 Chr.12: 122981668-122981668 on Build GRCh38
rs574457175 Chr.12: 122981703-122981703 on Build GRCh38
rs771692019 Chr.12: 122981818-122981818 on Build GRCh38
rs750242794 Chr.12: 122981720-122981720 on Build GRCh38
rs550025071 Chr.12: 122981760-122981760 on Build GRCh38
rs760355977 Chr.12: 122981823-122981823 on Build GRCh38
rs753633476 Chr.12: 122981832-122981832 on Build GRCh38
rs773573073 Chr.12: 122981692-122981692 on Build GRCh38
rs201538831 Chr.12: 122981612-122981612 on Build GRCh38
rs766730540 Chr.12: 122981637-122981637 on Build GRCh38
rs547482517 Chr.12: 122981798-122981798 on Build GRCh38
rs377049391 Chr.12: 122981685-122981685 on Build GRCh38
rs145515902 Chr.12: 122981781-122981781 on Build GRCh38
rs771311245 Chr.12: 122981698-122981698 on Build GRCh38
rs966204085 Chr.12: 122981712-122981712 on Build GRCh38
rs745875699 Chr.12: 122981746-122981748 on Build GRCh38
rs533949295 Chr.12: 122981749-122981749 on Build GRCh38
rs543285181 Chr.12: 122981697-122981697 on Build GRCh38
rs772478360 Chr.12: 122981615-122981615 on Build GRCh38
rs370149672 Chr.12: 122981711-122981711 on Build GRCh38
rs770718015 Chr.12: 122981746-122981754 on Build GRCh38
rs763803204 Chr.12: 122981751-122981752 on Build GRCh38
rs755931917 Chr.12: 122981721-122981721 on Build GRCh38
rs560155918 Chr.12: 122981722-122981722 on Build GRCh38
rs771239039 Chr.12: 122981771-122981771 on Build GRCh38
rs932720016 Chr.12: 122981683-122981683 on Build GRCh38
rs775081203 Chr.12: 122981640-122981640 on Build GRCh38
rs201978657 Chr.12: 122981679-122981679 on Build GRCh38
rs148921751 Chr.12: 122981649-122981649 on Build GRCh38
rs777030854 Chr.12: 122981782-122981782 on Build GRCh38
rs760473702 Chr.12: 122981748-122981748 on Build GRCh38
rs190753885 Chr.12: 122981643-122981643 on Build GRCh38
rs766169331 Chr.12: 122981733-122981733 on Build GRCh38
rs749115516 Chr.12: 122981740-122981742 on Build GRCh38
rs907004516 Chr.12: 122981661-122981661 on Build GRCh38
rs572450776 Chr.12: 122981620-122981620 on Build GRCh38
rs756079577 Chr.12: 122981671-122981671 on Build GRCh38
rs776802127 Chr.12: 122981753-122981757 on Build GRCh38
rs756947445 Chr.12: 122981655-122981655 on Build GRCh38
rs938565774 Chr.12: 122981785-122981785 on Build GRCh38
rs554208963 Chr.12: 122981616-122981616 on Build GRCh38
rs773998680 Chr.12: 122981797-122981797 on Build GRCh38
rs74758272 Chr.12: 122981757-122981757 on Build GRCh38
rs888534327 Chr.12: 122981683-122981684 on Build GRCh38
rs774335911 Chr.12: 122981746-122981751 on Build GRCh38
rs897731253 Chr.12: 122981721-122981721 on Build GRCh38
rs760277702 Chr.12: 122981750-122981750 on Build GRCh38
rs771545025 Chr.12: 122981745-122981746 on Build GRCh38
rs774625176 Chr.12: 122981602-122981602 on Build GRCh38
rs1037265957 Chr.12: 122981806-122981806 on Build GRCh38
rs977578175 Chr.12: 122981631-122981631 on Build GRCh38
rs755275821 Chr.12: 122981640-122981641 on Build GRCh38
rs566156092 Chr.12: 122981816-122981816 on Build GRCh38
rs772783141 Chr.12: 122981821-122981821 on Build GRCh38
rs760057114 Chr.12: 122981709-122981709 on Build GRCh38
rs767437870 Chr.12: 122981658-122981658 on Build GRCh38
rs543920304 Chr.12: 122981625-122981625 on Build GRCh38
rs763746548 Chr.12: 122981710-122981710 on Build GRCh38
rs186833977 Chr.12: 122981758-122981758 on Build GRCh38
rs1035757808 Chr.12: 122981740-122981740 on Build GRCh38
rs778995987 Chr.12: 122981752-122981752 on Build GRCh38
rs753772430 Chr.12: 122981743-122981743 on Build GRCh38
rs749247435 Chr.12: 122981677-122981677 on Build GRCh38
rs767208877 Chr.12: 122981717-122981717 on Build GRCh38
rs748395486 Chr.12: 122981686-122981686 on Build GRCh38
rs867776586 Chr.12: 122981817-122981817 on Build GRCh38
rs770381445 Chr.12: 122981792-122981792 on Build GRCh38
rs182360188 Chr.12: 122981724-122981724 on Build GRCh38

View Full Product Details