Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74066702-74066927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH6A1
Entrez Gene ID
4329
Gene Name
aldehyde dehydrogenase 6 family member A1
Gene Aliases
MMSADHA, MMSDH
UniGene
Hs.293970
Gene Chromosome Location
Chr.14: 74057662-74084493 on Build GRCh38
Transcripts
NM_001278593.1 NM_001278594.1 NM_005589.3 XM_017021331.1 XM_011537172.1 XM_017021659.1

Gene Symbol
BBOF1
Entrez Gene ID
80127
Gene Name
basal body orientation factor 1
Gene Aliases
C14orf45, CCDC176
UniGene
Hs.644621
Gene Chromosome Location
Chr.14: 74019350-74078351 on Build GRCh38
Transcripts
NM_001278593.1 NM_001278594.1 NM_005589.3 XM_017021331.1 XM_011537172.1 XM_017021659.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1582278 c.1139C>T p.S380F Chr.14: 74066790-74066790 on Build GRCh38
138942 c.1129G>A p.E377K Chr.14: 74066800-74066800 on Build GRCh38
4458349 c.1082C>T p.P361L Chr.14: 74066847-74066847 on Build GRCh38
6141296 c.1043G>A p.G348E Chr.14: 74066886-74066886 on Build GRCh38
7131152 c.1170G>A p.V390V Chr.14: 74066759-74066759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781767219 Chr.14: 74066773-74066773 on Build GRCh38
rs769625759 Chr.14: 74066745-74066745 on Build GRCh38
rs756777014 Chr.14: 74066833-74066833 on Build GRCh38
rs749236538 Chr.14: 74066762-74066762 on Build GRCh38
rs768541944 Chr.14: 74066764-74066764 on Build GRCh38
rs372922568 Chr.14: 74066772-74066772 on Build GRCh38
rs147813764 Chr.14: 74066875-74066875 on Build GRCh38
rs766867526 Chr.14: 74066778-74066778 on Build GRCh38
rs780772496 Chr.14: 74066844-74066844 on Build GRCh38
rs990299954 Chr.14: 74066785-74066785 on Build GRCh38
rs765805092 Chr.14: 74066796-74066796 on Build GRCh38
rs760746001 Chr.14: 74066777-74066777 on Build GRCh38
rs750093532 Chr.14: 74066846-74066846 on Build GRCh38
rs745706362 Chr.14: 74066737-74066737 on Build GRCh38
rs760147790 Chr.14: 74066790-74066790 on Build GRCh38
rs199699588 Chr.14: 74066839-74066839 on Build GRCh38
rs779756937 Chr.14: 74066754-74066754 on Build GRCh38
rs962711650 Chr.14: 74066865-74066865 on Build GRCh38
rs748743811 Chr.14: 74066869-74066869 on Build GRCh38
rs144401884 Chr.14: 74066872-74066872 on Build GRCh38
rs768631743 Chr.14: 74066895-74066895 on Build GRCh38
rs755705622 Chr.14: 74066862-74066862 on Build GRCh38
rs751076261 Chr.14: 74066813-74066813 on Build GRCh38
rs754201918 Chr.14: 74066786-74066786 on Build GRCh38
rs774455890 Chr.14: 74066766-74066766 on Build GRCh38
rs761933511 Chr.14: 74066770-74066770 on Build GRCh38

View Full Product Details