Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45801003-45801248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNTTIP1
Entrez Gene ID
116092
Gene Name
deoxynucleotidyltransferase, terminal, interacting protein 1
Gene Aliases
C20orf167, Tdif1, dJ447F3.4
UniGene
Hs.472852
Gene Chromosome Location
Chr.20: 45791937-45811427 on Build GRCh38
Transcripts
NM_052951.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5632388 c.391G>C p.D131H Chr.20: 45801092-45801092 on Build GRCh38
1483683 c.437T>A p.I146K Chr.20: 45801138-45801138 on Build GRCh38
3423674 c.384C>A p.L128L Chr.20: 45801085-45801085 on Build GRCh38
5057352 c.420C>A p.T140T Chr.20: 45801121-45801121 on Build GRCh38
3547087 c.397G>A p.E133K Chr.20: 45801098-45801098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936259536 Chr.20: 45801057-45801057 on Build GRCh38
rs1039629268 Chr.20: 45801047-45801047 on Build GRCh38
rs752097152 Chr.20: 45801175-45801178 on Build GRCh38
rs780494644 Chr.20: 45801173-45801173 on Build GRCh38
rs758904947 Chr.20: 45801163-45801163 on Build GRCh38
rs1053448215 Chr.20: 45801127-45801127 on Build GRCh38
rs1031929942 Chr.20: 45801189-45801189 on Build GRCh38
rs757900446 Chr.20: 45801190-45801190 on Build GRCh38
rs750976954 Chr.20: 45801160-45801160 on Build GRCh38
rs760974030 Chr.20: 45801049-45801049 on Build GRCh38
rs753839035 Chr.20: 45801097-45801102 on Build GRCh38
rs754892774 Chr.20: 45801153-45801166 on Build GRCh38
rs1021363854 Chr.20: 45801154-45801154 on Build GRCh38
rs765900526 Chr.20: 45801110-45801110 on Build GRCh38
rs1042188679 Chr.20: 45801223-45801223 on Build GRCh38
rs753989367 Chr.20: 45801060-45801060 on Build GRCh38
rs775108186 Chr.20: 45801036-45801036 on Build GRCh38
rs752142435 Chr.20: 45801162-45801162 on Build GRCh38
rs374568306 Chr.20: 45801179-45801179 on Build GRCh38
rs761977756 Chr.20: 45801099-45801099 on Build GRCh38
rs11697801 Chr.20: 45801176-45801176 on Build GRCh38
rs764867361 Chr.20: 45801168-45801174 on Build GRCh38
rs764199897 Chr.20: 45801055-45801055 on Build GRCh38
rs367687176 Chr.20: 45801175-45801175 on Build GRCh38
rs777952452 Chr.20: 45801113-45801113 on Build GRCh38
rs893514508 Chr.20: 45801170-45801170 on Build GRCh38
rs766264284 Chr.20: 45801057-45801057 on Build GRCh38

View Full Product Details