Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 579513-579738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSG
Entrez Gene ID
682
Gene Name
basigin (Ok blood group)
Gene Aliases
5F7, CD147, EMMPRIN, OK, TCSF
UniGene
Hs.501293
Gene Chromosome Location
Chr.19: 571277-583493 on Build GRCh38
Transcripts
NM_001322243.1 NM_001728.3 NM_198589.2 NM_198590.2 NM_198591.2 XM_017027173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6613174 c.436G>A p.V146I Chr.19: 579520-579520 on Build GRCh38
4419381 c.473C>T p.S158F Chr.19: 579557-579557 on Build GRCh38
4823393 c.429C>T p.F143F Chr.19: 579513-579513 on Build GRCh38
4823357 c.452C>G p.S151C Chr.19: 579536-579536 on Build GRCh38
4288081 c.461T>C p.L154P Chr.19: 579545-579545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551388840 Chr.19: 579680-579680 on Build GRCh38
rs368943609 Chr.19: 579681-579681 on Build GRCh38
rs769970655 Chr.19: 579711-579711 on Build GRCh38
rs375081858 Chr.19: 579543-579543 on Build GRCh38
rs774867811 Chr.19: 579628-579628 on Build GRCh38
rs140558438 Chr.19: 579567-579567 on Build GRCh38
rs777292458 Chr.19: 579612-579612 on Build GRCh38
rs776568185 Chr.19: 579707-579709 on Build GRCh38
rs534935683 Chr.19: 579717-579717 on Build GRCh38
rs1005985982 Chr.19: 579633-579633 on Build GRCh38
rs184596705 Chr.19: 579712-579712 on Build GRCh38
rs375962499 Chr.19: 579669-579669 on Build GRCh38
rs761944450 Chr.19: 579589-579589 on Build GRCh38
rs776176517 Chr.19: 579574-579574 on Build GRCh38
rs142283391 Chr.19: 579636-579636 on Build GRCh38
rs747324316 Chr.19: 579690-579690 on Build GRCh38
rs760264753 Chr.19: 579603-579605 on Build GRCh38
rs764956985 Chr.19: 579647-579647 on Build GRCh38
rs11551910 Chr.19: 579618-579618 on Build GRCh38
rs777154605 Chr.19: 579557-579557 on Build GRCh38
rs150004629 Chr.19: 579593-579593 on Build GRCh38
rs2229662 Chr.19: 579610-579610 on Build GRCh38
rs766215668 Chr.19: 579681-579718 on Build GRCh38
rs752283788 Chr.19: 579609-579609 on Build GRCh38
rs763628599 Chr.19: 579656-579656 on Build GRCh38
rs774708014 Chr.19: 579582-579582 on Build GRCh38
rs529058293 Chr.19: 579632-579632 on Build GRCh38
rs752515024 Chr.19: 579648-579648 on Build GRCh38
rs749753413 Chr.19: 579672-579672 on Build GRCh38
rs758033342 Chr.19: 579655-579655 on Build GRCh38
rs747857408 Chr.19: 579544-579544 on Build GRCh38
rs369274639 Chr.19: 579704-579704 on Build GRCh38
rs11551903 Chr.19: 579552-579552 on Build GRCh38
rs1012831574 Chr.19: 579599-579599 on Build GRCh38
rs751279949 Chr.19: 579665-579665 on Build GRCh38
rs112540819 Chr.19: 579700-579700 on Build GRCh38
rs769197832 Chr.19: 579581-579581 on Build GRCh38
rs766196523 Chr.19: 579603-579603 on Build GRCh38
rs12609013 Chr.19: 579713-579713 on Build GRCh38
rs369549553 Chr.19: 579634-579634 on Build GRCh38
rs868435068 Chr.19: 579557-579558 on Build GRCh38
rs12608994 Chr.19: 579699-579699 on Build GRCh38
rs770933792 Chr.19: 579696-579696 on Build GRCh38
rs749874305 Chr.19: 579624-579624 on Build GRCh38
rs14704 Chr.19: 579540-579540 on Build GRCh38
rs370866473 Chr.19: 579613-579613 on Build GRCh38
rs760619666 Chr.19: 579594-579594 on Build GRCh38
rs772536873 Chr.19: 579684-579684 on Build GRCh38
rs1023817527 Chr.19: 579608-579608 on Build GRCh38
rs11551905 Chr.19: 579585-579585 on Build GRCh38
rs780689888 Chr.19: 579616-579616 on Build GRCh38
rs531651313 Chr.19: 579615-579615 on Build GRCh38
rs534544911 Chr.19: 579580-579580 on Build GRCh38
rs113907190 Chr.19: 579658-579658 on Build GRCh38
rs748351801 Chr.19: 579629-579629 on Build GRCh38
rs747487226 Chr.19: 579571-579571 on Build GRCh38
rs753663735 Chr.19: 579538-579538 on Build GRCh38
rs767573364 Chr.19: 579590-579590 on Build GRCh38
rs999322753 Chr.19: 579546-579546 on Build GRCh38
rs755563121 Chr.19: 579673-579673 on Build GRCh38
rs113445094 Chr.19: 579659-579659 on Build GRCh38
rs746332382 Chr.19: 579570-579570 on Build GRCh38
rs759476275 Chr.19: 579640-579640 on Build GRCh38
rs773294258 Chr.19: 579686-579686 on Build GRCh38
rs756714868 Chr.19: 579668-579668 on Build GRCh38
rs763006257 Chr.19: 579718-579718 on Build GRCh38
rs61729580 Chr.19: 579630-579630 on Build GRCh38
rs369839342 Chr.19: 579549-579549 on Build GRCh38
rs759538134 Chr.19: 579707-579708 on Build GRCh38
rs1803202 Chr.19: 579627-579627 on Build GRCh38
rs1045237058 Chr.19: 579714-579714 on Build GRCh38
rs371359926 Chr.19: 579637-579637 on Build GRCh38
rs565826251 Chr.19: 579639-579639 on Build GRCh38
rs375132118 Chr.19: 579702-579702 on Build GRCh38
rs367570309 Chr.19: 579604-579604 on Build GRCh38

View Full Product Details