Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 58789369-58789614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICC1
Entrez Gene ID
80114
Gene Name
BicC family RNA binding protein 1
Gene Aliases
BICC, CYSRD
UniGene
Hs.100261
Gene Chromosome Location
Chr.10: 58512970-58831437 on Build GRCh38
Transcripts
NM_001080512.2 XM_005270169.4 XM_011540185.2 XM_011540188.2 XM_011540189.2 XM_011540190.2 XM_011540191.2 XM_017016677.1 XM_017016678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1348383 c.730C>T p.R244C Chr.10: 58789391-58789391 on Build GRCh38
77338 c.725A>C p.K242T Chr.10: 58789386-58789386 on Build GRCh38
1348384 c.790G>A p.V264M Chr.10: 58789451-58789451 on Build GRCh38
6066269 c.753T>A p.T251T Chr.10: 58789414-58789414 on Build GRCh38
1321282 c.716T>A p.V239E Chr.10: 58789377-58789377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766193556 Chr.10: 58789436-58789436 on Build GRCh38
rs747723883 Chr.10: 58789482-58789482 on Build GRCh38
rs201679654 Chr.10: 58789451-58789451 on Build GRCh38
rs199650571 Chr.10: 58789450-58789450 on Build GRCh38
rs771485499 Chr.10: 58789493-58789493 on Build GRCh38
rs576961494 Chr.10: 58789557-58789557 on Build GRCh38
rs748931836 Chr.10: 58789489-58789489 on Build GRCh38
rs755225045 Chr.10: 58789463-58789463 on Build GRCh38
rs777577817 Chr.10: 58789487-58789487 on Build GRCh38
rs557355549 Chr.10: 58789407-58789407 on Build GRCh38
rs754002090 Chr.10: 58789424-58789424 on Build GRCh38
rs558502138 Chr.10: 58789484-58789484 on Build GRCh38
rs748503091 Chr.10: 58789579-58789579 on Build GRCh38
rs942858590 Chr.10: 58789552-58789552 on Build GRCh38
rs1016652214 Chr.10: 58789442-58789442 on Build GRCh38
rs780941790 Chr.10: 58789476-58789476 on Build GRCh38
rs758248444 Chr.10: 58789431-58789431 on Build GRCh38
rs199893462 Chr.10: 58789415-58789415 on Build GRCh38
rs760709072 Chr.10: 58789398-58789398 on Build GRCh38
rs746172470 Chr.10: 58789506-58789506 on Build GRCh38
rs774743983 Chr.10: 58789502-58789502 on Build GRCh38
rs539897923 Chr.10: 58789467-58789467 on Build GRCh38

View Full Product Details