Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66496296-66496565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf46
Entrez Gene ID
254778
Gene Name
chromosome 8 open reading frame 46
Gene Aliases
-
UniGene
Hs.268869
Gene Chromosome Location
Chr.8: 66493256-66518524 on Build GRCh38
Transcripts
NM_152765.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6113578 c.123G>T p.K41N Chr.8: 66496489-66496489 on Build GRCh38
4765265 c.104_105insCTCAGCAC p.Q36fs*7 Chr.8: 66496470-66496471 on Build GRCh38
4842647 c.106C>G p.Q36E Chr.8: 66496472-66496472 on Build GRCh38
5746874 c.126+2T>C p.? Chr.8: 66496494-66496494 on Build GRCh38
6328545 c.71-4G>A p.? Chr.8: 66496433-66496433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542434937 Chr.8: 66496319-66496319 on Build GRCh38
rs74412410 Chr.8: 66496356-66496356 on Build GRCh38
rs201973579 Chr.8: 66496421-66496421 on Build GRCh38
rs754066059 Chr.8: 66496503-66496503 on Build GRCh38
rs897667968 Chr.8: 66496391-66496391 on Build GRCh38
rs750485775 Chr.8: 66496515-66496515 on Build GRCh38
rs747399587 Chr.8: 66496427-66496427 on Build GRCh38
rs544533516 Chr.8: 66496358-66496358 on Build GRCh38
rs780496799 Chr.8: 66496393-66496393 on Build GRCh38
rs977536422 Chr.8: 66496335-66496335 on Build GRCh38
rs765298052 Chr.8: 66496514-66496514 on Build GRCh38
rs536839970 Chr.8: 66496528-66496528 on Build GRCh38
rs755313842 Chr.8: 66496543-66496543 on Build GRCh38
rs555649987 Chr.8: 66496344-66496344 on Build GRCh38
rs983614257 Chr.8: 66496376-66496376 on Build GRCh38
rs916577813 Chr.8: 66496486-66496486 on Build GRCh38
rs551283828 Chr.8: 66496326-66496326 on Build GRCh38
rs759617356 Chr.8: 66496451-66496453 on Build GRCh38
rs758912424 Chr.8: 66496419-66496419 on Build GRCh38
rs373155514 Chr.8: 66496453-66496453 on Build GRCh38
rs867400560 Chr.8: 66496369-66496369 on Build GRCh38
rs767456234 Chr.8: 66496501-66496501 on Build GRCh38
rs369256469 Chr.8: 66496468-66496468 on Build GRCh38
rs768257600 Chr.8: 66496442-66496442 on Build GRCh38
rs186444216 Chr.8: 66496366-66496366 on Build GRCh38
rs758858896 Chr.8: 66496521-66496521 on Build GRCh38
rs1031848014 Chr.8: 66496343-66496343 on Build GRCh38
rs774238238 Chr.8: 66496480-66496480 on Build GRCh38
rs746385448 Chr.8: 66496413-66496413 on Build GRCh38
rs200750536 Chr.8: 66496492-66496492 on Build GRCh38
rs752102096 Chr.8: 66496531-66496531 on Build GRCh38
rs780866776 Chr.8: 66496444-66496444 on Build GRCh38
rs1012777250 Chr.8: 66496440-66496440 on Build GRCh38
rs940119816 Chr.8: 66496359-66496359 on Build GRCh38
rs963616793 Chr.8: 66496451-66496451 on Build GRCh38
rs977052439 Chr.8: 66496348-66496348 on Build GRCh38
rs893749572 Chr.8: 66496318-66496318 on Build GRCh38
rs771005102 Chr.8: 66496476-66496476 on Build GRCh38
rs772662854 Chr.8: 66496458-66496458 on Build GRCh38
rs778906378 Chr.8: 66496394-66496394 on Build GRCh38
rs921539568 Chr.8: 66496350-66496350 on Build GRCh38
rs761995781 Chr.8: 66496513-66496513 on Build GRCh38
rs571912647 Chr.8: 66496475-66496475 on Build GRCh38
rs746386249 Chr.8: 66496354-66496354 on Build GRCh38

View Full Product Details