Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218362706-218362943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATIP
Entrez Gene ID
375307
Gene Name
ciliogenesis associated TTC17 interacting protein
Gene Aliases
C2orf62
UniGene
Hs.645453
Gene Chromosome Location
Chr.2: 218356757-218368393 on Build GRCh38
Transcripts
NM_001320865.1 NM_198559.1 XM_005246539.4 XM_005246541.4 XM_011511148.2 XM_011511149.2 XM_011511150.2 XM_017004053.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3838580 c.518C>G p.S173* Chr.2: 218362790-218362790 on Build GRCh38
5039970 c.552G>A p.M184I Chr.2: 218362824-218362824 on Build GRCh38
5963494 c.466G>A p.V156M Chr.2: 218362738-218362738 on Build GRCh38
3838581 c.624G>A p.L208L Chr.2: 218362896-218362896 on Build GRCh38
7358494 c.476G>C p.G159A Chr.2: 218362748-218362748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780510430 Chr.2: 218362874-218362874 on Build GRCh38
rs778563062 Chr.2: 218362727-218362727 on Build GRCh38
rs747991967 Chr.2: 218362738-218362738 on Build GRCh38
rs770392502 Chr.2: 218362773-218362773 on Build GRCh38
rs766328855 Chr.2: 218362807-218362808 on Build GRCh38
rs200618385 Chr.2: 218362875-218362875 on Build GRCh38
rs763408598 Chr.2: 218362831-218362831 on Build GRCh38
rs756960889 Chr.2: 218362872-218362872 on Build GRCh38
rs561708956 Chr.2: 218362919-218362919 on Build GRCh38
rs779113500 Chr.2: 218362850-218362850 on Build GRCh38
rs762116934 Chr.2: 218362913-218362913 on Build GRCh38
rs777580747 Chr.2: 218362851-218362851 on Build GRCh38
rs754818348 Chr.2: 218362844-218362844 on Build GRCh38
rs199630251 Chr.2: 218362834-218362834 on Build GRCh38
rs1028839934 Chr.2: 218362762-218362762 on Build GRCh38
rs143024735 Chr.2: 218362901-218362901 on Build GRCh38
rs746347537 Chr.2: 218362806-218362806 on Build GRCh38
rs750576996 Chr.2: 218362837-218362837 on Build GRCh38
rs774693907 Chr.2: 218362833-218362833 on Build GRCh38
rs777422256 Chr.2: 218362757-218362757 on Build GRCh38
rs376259379 Chr.2: 218362824-218362824 on Build GRCh38
rs746655964 Chr.2: 218362855-218362855 on Build GRCh38
rs746709193 Chr.2: 218362758-218362758 on Build GRCh38
rs201316724 Chr.2: 218362750-218362750 on Build GRCh38
rs965828482 Chr.2: 218362737-218362737 on Build GRCh38
rs765398125 Chr.2: 218362909-218362909 on Build GRCh38
rs767869513 Chr.2: 218362917-218362917 on Build GRCh38
rs138318485 Chr.2: 218362876-218362876 on Build GRCh38
rs776271441 Chr.2: 218362799-218362799 on Build GRCh38
rs917793567 Chr.2: 218362860-218362860 on Build GRCh38
rs762840299 Chr.2: 218362772-218362772 on Build GRCh38
rs764364540 Chr.2: 218362832-218362832 on Build GRCh38
rs765228490 Chr.2: 218362846-218362846 on Build GRCh38
rs140310382 Chr.2: 218362752-218362752 on Build GRCh38
rs757312383 Chr.2: 218362840-218362840 on Build GRCh38
rs145903057 Chr.2: 218362808-218362808 on Build GRCh38
rs776089397 Chr.2: 218362877-218362877 on Build GRCh38
rs201618489 Chr.2: 218362835-218362835 on Build GRCh38
rs752383249 Chr.2: 218362848-218362848 on Build GRCh38
rs749861660 Chr.2: 218362887-218362887 on Build GRCh38
rs758164600 Chr.2: 218362849-218362849 on Build GRCh38
rs984536720 Chr.2: 218362797-218362797 on Build GRCh38
rs774923000 Chr.2: 218362902-218362902 on Build GRCh38

View Full Product Details