Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 149933419-149933690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf40B
Entrez Gene ID
541578
Gene Name
chromosome X open reading frame 40B
Gene Aliases
-
UniGene
Hs.545950
Gene Chromosome Location
Chr.X: 149924152-149938850 on Build GRCh38
Transcripts
NM_001013845.1 XM_005274698.1 XM_005274699.1 XM_005274700.1 XM_005274701.2 XM_005274702.3 XM_006724826.3 XM_011531180.2 XM_011531181.2 XM_017029585.1 XM_017029586.1 XM_017029587.1 XM_017029588.1 XM_017029589.1 XM_017029590.1 XM_017029591.1 XM_017029592.1 XM_017029593.1

SNP Detail

SNP ID Chromosome Location
rs369727000 Chr.X: 149933637-149933637 on Build GRCh38
rs782286243 Chr.X: 149933641-149933641 on Build GRCh38
rs782190549 Chr.X: 149933594-149933594 on Build GRCh38
rs782637156 Chr.X: 149933588-149933588 on Build GRCh38
rs782552724 Chr.X: 149933585-149933585 on Build GRCh38
rs782644545 Chr.X: 149933649-149933649 on Build GRCh38
rs781806879 Chr.X: 149933618-149933618 on Build GRCh38
rs782284591 Chr.X: 149933582-149933582 on Build GRCh38
rs781785257 Chr.X: 149933624-149933624 on Build GRCh38
rs372285759 Chr.X: 149933590-149933590 on Build GRCh38
rs782400138 Chr.X: 149933610-149933610 on Build GRCh38
rs782588985 Chr.X: 149933579-149933579 on Build GRCh38
rs782223399 Chr.X: 149933653-149933653 on Build GRCh38
rs782102073 Chr.X: 149933611-149933611 on Build GRCh38
rs781787409 Chr.X: 149933635-149933635 on Build GRCh38
rs781842553 Chr.X: 149933575-149933575 on Build GRCh38
rs782618058 Chr.X: 149933633-149933633 on Build GRCh38
rs782367794 Chr.X: 149933655-149933655 on Build GRCh38
rs201379178 Chr.X: 149933598-149933598 on Build GRCh38
rs782456006 Chr.X: 149933576-149933576 on Build GRCh38
rs782111872 Chr.X: 149933599-149933599 on Build GRCh38
rs782696178 Chr.X: 149933613-149933613 on Build GRCh38
rs782146551 Chr.X: 149933621-149933621 on Build GRCh38
rs781915018 Chr.X: 149933592-149933592 on Build GRCh38
rs782368609 Chr.X: 149933644-149933644 on Build GRCh38
rs782467805 Chr.X: 149933631-149933631 on Build GRCh38
rs782069729 Chr.X: 149933665-149933665 on Build GRCh38
rs781872415 Chr.X: 149933477-149933477 on Build GRCh38
rs782811422 Chr.X: 149933623-149933623 on Build GRCh38
rs782208761 Chr.X: 149933589-149933589 on Build GRCh38
rs782464013 Chr.X: 149933636-149933636 on Build GRCh38
rs782751096 Chr.X: 149933571-149933571 on Build GRCh38
rs781923135 Chr.X: 149933659-149933659 on Build GRCh38
rs782534614 Chr.X: 149933513-149933513 on Build GRCh38

View Full Product Details