Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68822270-68822529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP25
Entrez Gene ID
9938
Gene Name
Rho GTPase activating protein 25
Gene Aliases
HEL-S-308, KAIA0053
UniGene
Hs.531807
Gene Chromosome Location
Chr.2: 68710505-68826833 on Build GRCh38
Transcripts
NM_001007231.2 NM_001166276.1 NM_001166277.1 NM_014882.2 XM_005264675.2 XM_005264676.2 XM_011533206.2 XM_011533207.2 XM_011533208.2 XM_011533209.2 XM_011533210.2 XM_017005423.1 XM_017005424.1 XM_017005425.1 XM_017005426.1 XM_017005427.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6092104 c.1359C>A p.S453R Chr.2: 68822519-68822519 on Build GRCh38
5983684 c.1350C>G p.F450L Chr.2: 68822489-68822489 on Build GRCh38
1641957 c.1202G>A p.S401N Chr.2: 68822362-68822362 on Build GRCh38
5354274 c.1245C>T p.S415S Chr.2: 68822384-68822384 on Build GRCh38
3770725 c.1227T>C p.D409D Chr.2: 68822387-68822387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375050207 Chr.2: 68822384-68822384 on Build GRCh38
rs150901180 Chr.2: 68822406-68822406 on Build GRCh38
rs758811327 Chr.2: 68822494-68822494 on Build GRCh38
rs1027320647 Chr.2: 68822424-68822424 on Build GRCh38
rs141322544 Chr.2: 68822355-68822355 on Build GRCh38
rs756773730 Chr.2: 68822370-68822370 on Build GRCh38
rs762774150 Chr.2: 68822417-68822417 on Build GRCh38
rs774443296 Chr.2: 68822322-68822322 on Build GRCh38
rs752414071 Chr.2: 68822354-68822354 on Build GRCh38
rs748691934 Chr.2: 68822490-68822490 on Build GRCh38
rs1039588541 Chr.2: 68822405-68822405 on Build GRCh38
rs765122338 Chr.2: 68822353-68822353 on Build GRCh38
rs747107253 Chr.2: 68822348-68822349 on Build GRCh38
rs201588014 Chr.2: 68822441-68822441 on Build GRCh38
rs761008026 Chr.2: 68822404-68822404 on Build GRCh38
rs760801423 Chr.2: 68822350-68822350 on Build GRCh38
rs143476860 Chr.2: 68822390-68822390 on Build GRCh38
rs146486959 Chr.2: 68822369-68822369 on Build GRCh38
rs948164561 Chr.2: 68822340-68822340 on Build GRCh38
rs749851241 Chr.2: 68822455-68822455 on Build GRCh38
rs150035391 Chr.2: 68822483-68822483 on Build GRCh38
rs769098140 Chr.2: 68822319-68822319 on Build GRCh38
rs150157837 Chr.2: 68822376-68822376 on Build GRCh38
rs565639760 Chr.2: 68822407-68822407 on Build GRCh38
rs140270836 Chr.2: 68822396-68822396 on Build GRCh38
rs370858973 Chr.2: 68822381-68822381 on Build GRCh38
rs765877536 Chr.2: 68822478-68822478 on Build GRCh38
rs200609383 Chr.2: 68822479-68822479 on Build GRCh38
rs767100810 Chr.2: 68822451-68822451 on Build GRCh38
rs773539945 Chr.2: 68822349-68822349 on Build GRCh38
rs373864170 Chr.2: 68822311-68822311 on Build GRCh38
rs761814522 Chr.2: 68822336-68822336 on Build GRCh38
rs755818506 Chr.2: 68822468-68822468 on Build GRCh38
rs777933931 Chr.2: 68822506-68822506 on Build GRCh38
rs780247143 Chr.2: 68822305-68822305 on Build GRCh38
rs764144032 Chr.2: 68822429-68822429 on Build GRCh38
rs772295238 Chr.2: 68822345-68822345 on Build GRCh38
rs376461540 Chr.2: 68822395-68822395 on Build GRCh38
rs143574274 Chr.2: 68822387-68822387 on Build GRCh38
rs374170016 Chr.2: 68822485-68822485 on Build GRCh38
rs928014320 Chr.2: 68822359-68822359 on Build GRCh38
rs780557448 Chr.2: 68822380-68822380 on Build GRCh38
rs764084464 Chr.2: 68822365-68822365 on Build GRCh38
rs941277487 Chr.2: 68822400-68822400 on Build GRCh38
rs761840762 Chr.2: 68822447-68822447 on Build GRCh38
rs149291667 Chr.2: 68822385-68822385 on Build GRCh38
rs773134106 Chr.2: 68822399-68822399 on Build GRCh38

View Full Product Details