Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138209730-138209957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC8
Entrez Gene ID
25852
Gene Name
armadillo repeat containing 8
Gene Aliases
GID5, HSPC056, S863-2, VID28
UniGene
Hs.744868
Gene Chromosome Location
Chr.3: 138187248-138298389 on Build GRCh38
Transcripts
NM_001267041.2 NM_001267042.2 NM_001282342.1 NM_014154.4 NM_015396.5 NM_213654.3 XM_006713564.3 XM_006713565.2 XM_006713566.3 XM_006713567.3 XM_011512632.1 XM_011512633.1 XM_017006095.1 XM_017006096.1 XM_017006097.1 XM_017006098.1 XM_017006099.1 XM_017006100.1 XM_017006101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
204941 c.23G>A p.R8H Chr.3: 138209836-138209836 on Build GRCh38
6298521 c.80+1G>T p.? Chr.3: 138209894-138209894 on Build GRCh38
3123845 c.13G>A p.A5T Chr.3: 138209826-138209826 on Build GRCh38
5742996 c.55G>A p.A19T Chr.3: 138209826-138209826 on Build GRCh38
204940 c.23G>A p.R8H Chr.3: 138209836-138209836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755439802 Chr.3: 138209921-138209921 on Build GRCh38
rs765325711 Chr.3: 138209878-138209878 on Build GRCh38
rs997131922 Chr.3: 138209873-138209873 on Build GRCh38
rs781753477 Chr.3: 138209801-138209801 on Build GRCh38
rs763780226 Chr.3: 138209855-138209855 on Build GRCh38
rs776613707 Chr.3: 138209858-138209858 on Build GRCh38
rs1040509518 Chr.3: 138209807-138209807 on Build GRCh38
rs377205828 Chr.3: 138209905-138209905 on Build GRCh38
rs200908758 Chr.3: 138209877-138209877 on Build GRCh38
rs758574873 Chr.3: 138209769-138209769 on Build GRCh38
rs192366102 Chr.3: 138209808-138209808 on Build GRCh38
rs780318301 Chr.3: 138209774-138209774 on Build GRCh38
rs773974549 Chr.3: 138209814-138209814 on Build GRCh38
rs200907842 Chr.3: 138209828-138209828 on Build GRCh38
rs372678205 Chr.3: 138209783-138209783 on Build GRCh38
rs771715573 Chr.3: 138209833-138209833 on Build GRCh38
rs770604420 Chr.3: 138209809-138209809 on Build GRCh38
rs753161864 Chr.3: 138209928-138209931 on Build GRCh38
rs372967231 Chr.3: 138209842-138209842 on Build GRCh38
rs370126943 Chr.3: 138209919-138209919 on Build GRCh38
rs758627941 Chr.3: 138209907-138209907 on Build GRCh38
rs767569152 Chr.3: 138209918-138209919 on Build GRCh38
rs769057888 Chr.3: 138209793-138209793 on Build GRCh38
rs908966744 Chr.3: 138209796-138209797 on Build GRCh38
rs759604518 Chr.3: 138209792-138209792 on Build GRCh38

View Full Product Details