Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 12824396-12824659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK1D
Entrez Gene ID
57118
Gene Name
calcium/calmodulin dependent protein kinase ID
Gene Aliases
CKLiK, CaM-K1, CaMKID
UniGene
Hs.600547
Gene Chromosome Location
Chr.10: 12349514-12835545 on Build GRCh38
Transcripts
NM_020397.3 NM_153498.3 XM_006717481.3 XM_006717482.3 XM_006717483.3 XM_011519591.2 XM_011519592.2 XM_011519593.2 XM_011519594.2 XM_011519595.2 XM_017016438.1 XM_017016439.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6334012 c.911G>T p.S304I Chr.10: 12824542-12824542 on Build GRCh38
4637243 c.906C>T p.A302A Chr.10: 12824537-12824537 on Build GRCh38
6225074 c.912C>T p.S304S Chr.10: 12824543-12824543 on Build GRCh38
6334011 c.911G>T p.S304I Chr.10: 12824542-12824542 on Build GRCh38
4637244 c.906C>T p.A302A Chr.10: 12824537-12824537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201511697 Chr.10: 12824594-12824594 on Build GRCh38
rs977199993 Chr.10: 12824439-12824439 on Build GRCh38
rs763045698 Chr.10: 12824591-12824591 on Build GRCh38
rs138858875 Chr.10: 12824499-12824499 on Build GRCh38
rs750385169 Chr.10: 12824514-12824514 on Build GRCh38
rs916157316 Chr.10: 12824495-12824495 on Build GRCh38
rs912179319 Chr.10: 12824469-12824469 on Build GRCh38
rs759652179 Chr.10: 12824447-12824447 on Build GRCh38
rs372991251 Chr.10: 12824423-12824423 on Build GRCh38
rs200929443 Chr.10: 12824513-12824513 on Build GRCh38
rs376511947 Chr.10: 12824580-12824580 on Build GRCh38
rs191189221 Chr.10: 12824562-12824562 on Build GRCh38
rs889020807 Chr.10: 12824597-12824597 on Build GRCh38
rs200034782 Chr.10: 12824468-12824468 on Build GRCh38
rs761771196 Chr.10: 12824595-12824595 on Build GRCh38
rs371723953 Chr.10: 12824420-12824420 on Build GRCh38
rs748766971 Chr.10: 12824551-12824551 on Build GRCh38
rs777098698 Chr.10: 12824444-12824444 on Build GRCh38
rs201893185 Chr.10: 12824524-12824524 on Build GRCh38
rs142495899 Chr.10: 12824477-12824477 on Build GRCh38
rs764275569 Chr.10: 12824593-12824593 on Build GRCh38
rs770274557 Chr.10: 12824579-12824579 on Build GRCh38
rs373324078 Chr.10: 12824570-12824570 on Build GRCh38
rs746539488 Chr.10: 12824447-12824448 on Build GRCh38
rs767411075 Chr.10: 12824596-12824596 on Build GRCh38
rs768226034 Chr.10: 12824555-12824555 on Build GRCh38
rs764307882 Chr.10: 12824501-12824501 on Build GRCh38
rs760989056 Chr.10: 12824434-12824434 on Build GRCh38
rs757099962 Chr.10: 12824508-12824508 on Build GRCh38
rs991717263 Chr.10: 12824488-12824488 on Build GRCh38
rs760019921 Chr.10: 12824578-12824578 on Build GRCh38
rs747421667 Chr.10: 12824568-12824568 on Build GRCh38
rs369005523 Chr.10: 12824451-12824451 on Build GRCh38
rs572328546 Chr.10: 12824576-12824576 on Build GRCh38
rs145937671 Chr.10: 12824507-12824507 on Build GRCh38
rs375709397 Chr.10: 12824421-12824421 on Build GRCh38
rs779443327 Chr.10: 12824528-12824528 on Build GRCh38
rs766893329 Chr.10: 12824442-12824442 on Build GRCh38

View Full Product Details