Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96761326-96761597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM4
Entrez Gene ID
26504
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 4
Gene Aliases
ACDP4
UniGene
Hs.175043
Gene Chromosome Location
Chr.2: 96760902-96811891 on Build GRCh38
Transcripts
NM_020184.3 XM_005263914.3 XM_005263915.3 XM_011510955.2 XM_011510956.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6631346 c.559C>A p.L187I Chr.2: 96761558-96761558 on Build GRCh38
6631344 c.502T>G p.S168A Chr.2: 96761501-96761501 on Build GRCh38
1494997 c.391G>A p.V131M Chr.2: 96761390-96761390 on Build GRCh38
6234650 c.411G>T p.K137N Chr.2: 96761410-96761410 on Build GRCh38
3910802 c.527C>T p.P176L Chr.2: 96761526-96761526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764182054 Chr.2: 96761533-96761533 on Build GRCh38
rs767683968 Chr.2: 96761542-96761542 on Build GRCh38
rs755932169 Chr.2: 96761548-96761548 on Build GRCh38
rs373487333 Chr.2: 96761532-96761532 on Build GRCh38
rs532668909 Chr.2: 96761473-96761473 on Build GRCh38
rs977161267 Chr.2: 96761401-96761401 on Build GRCh38
rs368862739 Chr.2: 96761348-96761348 on Build GRCh38
rs34147094 Chr.2: 96761398-96761398 on Build GRCh38
rs377354317 Chr.2: 96761362-96761362 on Build GRCh38
rs745841697 Chr.2: 96761347-96761347 on Build GRCh38
rs770328147 Chr.2: 96761500-96761500 on Build GRCh38
rs751741858 Chr.2: 96761539-96761539 on Build GRCh38
rs760053491 Chr.2: 96761404-96761404 on Build GRCh38
rs778099516 Chr.2: 96761468-96761481 on Build GRCh38
rs764233273 Chr.2: 96761416-96761416 on Build GRCh38
rs752806783 Chr.2: 96761409-96761409 on Build GRCh38
rs144179610 Chr.2: 96761553-96761553 on Build GRCh38
rs1027643631 Chr.2: 96761560-96761560 on Build GRCh38
rs766867656 Chr.2: 96761386-96761386 on Build GRCh38
rs370486600 Chr.2: 96761428-96761428 on Build GRCh38
rs748830242 Chr.2: 96761447-96761447 on Build GRCh38
rs761058012 Chr.2: 96761380-96761380 on Build GRCh38
rs552814825 Chr.2: 96761547-96761547 on Build GRCh38
rs370665884 Chr.2: 96761387-96761387 on Build GRCh38
rs560847070 Chr.2: 96761471-96761471 on Build GRCh38
rs17855817 Chr.2: 96761375-96761375 on Build GRCh38
rs775036266 Chr.2: 96761352-96761352 on Build GRCh38
rs148129550 Chr.2: 96761433-96761433 on Build GRCh38
rs778656128 Chr.2: 96761464-96761464 on Build GRCh38
rs777028877 Chr.2: 96761474-96761474 on Build GRCh38
rs561036245 Chr.2: 96761364-96761364 on Build GRCh38
rs759874330 Chr.2: 96761484-96761484 on Build GRCh38
rs745779788 Chr.2: 96761402-96761402 on Build GRCh38
rs889281402 Chr.2: 96761439-96761439 on Build GRCh38
rs146760430 Chr.2: 96761555-96761555 on Build GRCh38
rs376702001 Chr.2: 96761541-96761541 on Build GRCh38
rs768413195 Chr.2: 96761359-96761359 on Build GRCh38
rs897829700 Chr.2: 96761552-96761552 on Build GRCh38
rs377110822 Chr.2: 96761422-96761422 on Build GRCh38
rs149951530 Chr.2: 96761569-96761569 on Build GRCh38
rs999429293 Chr.2: 96761360-96761360 on Build GRCh38
rs945013257 Chr.2: 96761507-96761507 on Build GRCh38
rs765710051 Chr.2: 96761406-96761406 on Build GRCh38
rs754494081 Chr.2: 96761563-96761563 on Build GRCh38
rs779915981 Chr.2: 96761446-96761446 on Build GRCh38
rs756997497 Chr.2: 96761417-96761417 on Build GRCh38
rs775865145 Chr.2: 96761516-96761516 on Build GRCh38
rs768290533 Chr.2: 96761457-96761457 on Build GRCh38
rs758539045 Chr.2: 96761410-96761410 on Build GRCh38

View Full Product Details