Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169424746-169425017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC181
Entrez Gene ID
57821
Gene Name
coiled-coil domain containing 181
Gene Aliases
C1orf114
UniGene
Hs.567557
Gene Chromosome Location
Chr.1: 169394870-169462221 on Build GRCh38
Transcripts
NM_001300968.1 NM_001300969.1 NM_021179.2 XM_005245383.1 XM_011509827.2 XM_017001938.1 XM_017001939.1 XM_017001940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1668199 c.67C>A p.L23M Chr.1: 169424861-169424861 on Build GRCh38
3705221 c.16G>A p.D6N Chr.1: 169424912-169424912 on Build GRCh38
3477956 c.91G>A p.E31K Chr.1: 169424837-169424837 on Build GRCh38
6122304 c.58G>C p.E20Q Chr.1: 169424870-169424870 on Build GRCh38
4024913 c.92A>G p.E31G Chr.1: 169424836-169424836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951753422 Chr.1: 169424959-169424959 on Build GRCh38
rs774233968 Chr.1: 169424895-169424895 on Build GRCh38
rs144603502 Chr.1: 169424837-169424837 on Build GRCh38
rs199771563 Chr.1: 169424883-169424883 on Build GRCh38
rs149997629 Chr.1: 169424819-169424819 on Build GRCh38
rs759171287 Chr.1: 169424820-169424820 on Build GRCh38
rs377120788 Chr.1: 169424788-169424788 on Build GRCh38
rs775165865 Chr.1: 169424946-169424946 on Build GRCh38
rs199660251 Chr.1: 169424924-169424924 on Build GRCh38
rs750936997 Chr.1: 169424957-169424957 on Build GRCh38
rs755125820 Chr.1: 169424879-169424879 on Build GRCh38
rs760442951 Chr.1: 169424921-169424922 on Build GRCh38
rs760241695 Chr.1: 169424950-169424950 on Build GRCh38
rs751918681 Chr.1: 169424905-169424905 on Build GRCh38
rs200927034 Chr.1: 169424845-169424845 on Build GRCh38
rs780208241 Chr.1: 169424862-169424862 on Build GRCh38
rs759270486 Chr.1: 169424917-169424917 on Build GRCh38
rs765985395 Chr.1: 169424811-169424811 on Build GRCh38
rs758760085 Chr.1: 169424858-169424858 on Build GRCh38
rs752192458 Chr.1: 169424840-169424840 on Build GRCh38
rs369306577 Chr.1: 169424847-169424847 on Build GRCh38
rs773167593 Chr.1: 169424886-169424886 on Build GRCh38
rs369278363 Chr.1: 169424977-169424977 on Build GRCh38
rs770742782 Chr.1: 169424892-169424892 on Build GRCh38
rs201674076 Chr.1: 169424863-169424863 on Build GRCh38
rs372753443 Chr.1: 169424978-169424978 on Build GRCh38
rs567643312 Chr.1: 169424979-169424979 on Build GRCh38
rs765705799 Chr.1: 169424951-169424951 on Build GRCh38
rs549609366 Chr.1: 169424967-169424967 on Build GRCh38
rs544824549 Chr.1: 169424880-169424880 on Build GRCh38
rs995676702 Chr.1: 169424933-169424933 on Build GRCh38
rs766770009 Chr.1: 169424968-169424968 on Build GRCh38
rs375735868 Chr.1: 169424882-169424882 on Build GRCh38
rs749100027 Chr.1: 169424891-169424891 on Build GRCh38
rs762783597 Chr.1: 169424807-169424807 on Build GRCh38
rs10919168 Chr.1: 169424988-169424988 on Build GRCh38
rs370211562 Chr.1: 169424790-169424790 on Build GRCh38
rs974661044 Chr.1: 169424991-169424991 on Build GRCh38
rs772903383 Chr.1: 169424806-169424806 on Build GRCh38
rs767017590 Chr.1: 169424856-169424856 on Build GRCh38
rs775306099 Chr.1: 169424844-169424847 on Build GRCh38

View Full Product Details