Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 75053311-75053582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB7
Entrez Gene ID
22
Gene Name
ATP binding cassette subfamily B member 7
Gene Aliases
ABC7, ASAT, Atm1p, EST140535
UniGene
Hs.370480
Gene Chromosome Location
Chr.X: 75053172-75156340 on Build GRCh38
Transcripts
NM_001271696.1 NM_001271697.1 NM_001271698.1 NM_001271699.1 NM_004299.4

SNP Detail

SNP ID Chromosome Location
rs917894774 Chr.X: 75053451-75053451 on Build GRCh38
rs757771281 Chr.X: 75053539-75053539 on Build GRCh38
rs758530677 Chr.X: 75053478-75053478 on Build GRCh38
rs771854345 Chr.X: 75053376-75053376 on Build GRCh38
rs748936726 Chr.X: 75053347-75053347 on Build GRCh38
rs752113839 Chr.X: 75053522-75053522 on Build GRCh38
rs781524899 Chr.X: 75053556-75053556 on Build GRCh38
rs760317782 Chr.X: 75053400-75053400 on Build GRCh38
rs755815706 Chr.X: 75053362-75053363 on Build GRCh38
rs768257406 Chr.X: 75053563-75053563 on Build GRCh38
rs863223869 Chr.X: 75053443-75053443 on Build GRCh38
rs751986553 Chr.X: 75053353-75053353 on Build GRCh38
rs761416644 Chr.X: 75053355-75053355 on Build GRCh38
rs201924920 Chr.X: 75053419-75053419 on Build GRCh38
rs752787356 Chr.X: 75053477-75053477 on Build GRCh38
rs949445106 Chr.X: 75053497-75053497 on Build GRCh38
rs746271411 Chr.X: 75053559-75053559 on Build GRCh38
rs768355588 Chr.X: 75053348-75053348 on Build GRCh38
rs372891558 Chr.X: 75053377-75053377 on Build GRCh38
rs765601526 Chr.X: 75053450-75053450 on Build GRCh38
rs371035361 Chr.X: 75053423-75053423 on Build GRCh38
rs377609329 Chr.X: 75053404-75053404 on Build GRCh38
rs750002950 Chr.X: 75053344-75053345 on Build GRCh38
rs1045718568 Chr.X: 75053533-75053533 on Build GRCh38
rs778085122 Chr.X: 75053485-75053485 on Build GRCh38
rs866082104 Chr.X: 75053456-75053456 on Build GRCh38

View Full Product Details