Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 75071424-75071663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB7
Entrez Gene ID
22
Gene Name
ATP binding cassette subfamily B member 7
Gene Aliases
ABC7, ASAT, Atm1p, EST140535
UniGene
Hs.370480
Gene Chromosome Location
Chr.X: 75053172-75156340 on Build GRCh38
Transcripts
NM_001271696.1 NM_001271697.1 NM_001271698.1 NM_001271699.1 NM_004299.4

SNP Detail

SNP ID Chromosome Location
rs752639475 Chr.X: 75071501-75071501 on Build GRCh38
rs756266541 Chr.X: 75071555-75071555 on Build GRCh38
rs72554634 Chr.X: 75071516-75071516 on Build GRCh38
rs758400043 Chr.X: 75071507-75071507 on Build GRCh38
rs749798732 Chr.X: 75071585-75071585 on Build GRCh38
rs1052386741 Chr.X: 75071517-75071517 on Build GRCh38
rs955046524 Chr.X: 75071632-75071632 on Build GRCh38
rs139311518 Chr.X: 75071542-75071542 on Build GRCh38
rs201269113 Chr.X: 75071484-75071484 on Build GRCh38
rs1008068103 Chr.X: 75071586-75071586 on Build GRCh38
rs887974659 Chr.X: 75071491-75071491 on Build GRCh38
rs1029747021 Chr.X: 75071631-75071631 on Build GRCh38
rs962282106 Chr.X: 75071614-75071614 on Build GRCh38
rs780346552 Chr.X: 75071566-75071566 on Build GRCh38
rs778824146 Chr.X: 75071466-75071466 on Build GRCh38
rs769334662 Chr.X: 75071630-75071630 on Build GRCh38
rs765098755 Chr.X: 75071499-75071499 on Build GRCh38
rs751420864 Chr.X: 75071515-75071515 on Build GRCh38
rs763867013 Chr.X: 75071513-75071513 on Build GRCh38

View Full Product Details