Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67979572-67979835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPEP3
Entrez Gene ID
64180
Gene Name
dipeptidase 3
Gene Aliases
MBD3
UniGene
Hs.302028
Gene Chromosome Location
Chr.16: 67975663-67980549 on Build GRCh38
Transcripts
NM_001129758.1 NM_022357.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7295661 c.376C>T p.P126S Chr.16: 67979752-67979752 on Build GRCh38
6369941 c.484G>T p.A162S Chr.16: 67979644-67979644 on Build GRCh38
7320586 c.447G>C p.Q149H Chr.16: 67979681-67979681 on Build GRCh38
1709309 c.473G>A p.G158D Chr.16: 67979655-67979655 on Build GRCh38
5959684 c.482G>T p.G161V Chr.16: 67979646-67979646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766972461 Chr.16: 67979729-67979729 on Build GRCh38
rs7189697 Chr.16: 67979656-67979656 on Build GRCh38
rs753163516 Chr.16: 67979810-67979810 on Build GRCh38
rs529760037 Chr.16: 67979638-67979638 on Build GRCh38
rs201060618 Chr.16: 67979607-67979607 on Build GRCh38
rs749228204 Chr.16: 67979771-67979771 on Build GRCh38
rs527620672 Chr.16: 67979660-67979660 on Build GRCh38
rs776648183 Chr.16: 67979605-67979605 on Build GRCh38
rs777133028 Chr.16: 67979789-67979789 on Build GRCh38
rs760286403 Chr.16: 67979783-67979783 on Build GRCh38
rs370294992 Chr.16: 67979761-67979761 on Build GRCh38
rs535894089 Chr.16: 67979773-67979773 on Build GRCh38
rs755341288 Chr.16: 67979734-67979734 on Build GRCh38
rs765704998 Chr.16: 67979735-67979735 on Build GRCh38
rs777695414 Chr.16: 67979760-67979760 on Build GRCh38
rs767020082 Chr.16: 67979646-67979646 on Build GRCh38
rs61743289 Chr.16: 67979651-67979651 on Build GRCh38
rs1014861788 Chr.16: 67979726-67979726 on Build GRCh38
rs200922436 Chr.16: 67979667-67979667 on Build GRCh38
rs768398646 Chr.16: 67979772-67979772 on Build GRCh38
rs769432385 Chr.16: 67979608-67979608 on Build GRCh38
rs752930586 Chr.16: 67979744-67979744 on Build GRCh38
rs550111634 Chr.16: 67979759-67979759 on Build GRCh38
rs770356228 Chr.16: 67979784-67979784 on Build GRCh38
rs200788983 Chr.16: 67979689-67979689 on Build GRCh38
rs772825295 Chr.16: 67979721-67979721 on Build GRCh38
rs35185915 Chr.16: 67979594-67979595 on Build GRCh38
rs56921747 Chr.16: 67979690-67979690 on Build GRCh38
rs777936412 Chr.16: 67979678-67979678 on Build GRCh38
rs763775016 Chr.16: 67979628-67979628 on Build GRCh38
rs749174219 Chr.16: 67979683-67979683 on Build GRCh38
rs760155804 Chr.16: 67979795-67979795 on Build GRCh38
rs772589313 Chr.16: 67979776-67979776 on Build GRCh38
rs766963169 Chr.16: 67979713-67979713 on Build GRCh38
rs745741301 Chr.16: 67979679-67979679 on Build GRCh38
rs754135725 Chr.16: 67979733-67979733 on Build GRCh38
rs541668928 Chr.16: 67979650-67979650 on Build GRCh38
rs112171888 Chr.16: 67979623-67979623 on Build GRCh38
rs368791692 Chr.16: 67979647-67979647 on Build GRCh38
rs760152097 Chr.16: 67979809-67979818 on Build GRCh38
rs752047736 Chr.16: 67979768-67979768 on Build GRCh38
rs775375923 Chr.16: 67979615-67979615 on Build GRCh38
rs771795286 Chr.16: 67979774-67979774 on Build GRCh38
rs765651671 Chr.16: 67979796-67979796 on Build GRCh38
rs139520527 Chr.16: 67979681-67979681 on Build GRCh38
rs762490078 Chr.16: 67979626-67979626 on Build GRCh38
rs780098137 Chr.16: 67979768-67979768 on Build GRCh38
rs769817080 Chr.16: 67979680-67979680 on Build GRCh38
rs773926731 Chr.16: 67979685-67979685 on Build GRCh38
rs761053221 Chr.16: 67979722-67979722 on Build GRCh38
rs768284113 Chr.16: 67979684-67979684 on Build GRCh38
rs757805500 Chr.16: 67979637-67979637 on Build GRCh38

View Full Product Details