Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49718309-49718578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMIGO3
Entrez Gene ID
386724
Gene Name
adhesion molecule with Ig like domain 3
Gene Aliases
ALI3, AMIGO-3
UniGene
Hs.567903
Gene Chromosome Location
Chr.3: 49716834-49719805 on Build GRCh38
Transcripts
NM_198722.2 NM_022064.4 NR_135218.1 XM_006713285.1 XM_011533995.1 XM_017007018.1

Gene Symbol
RNF123
Entrez Gene ID
63891
Gene Name
ring finger protein 123
Gene Aliases
FP1477, KPC1
UniGene
Hs.553723
Gene Chromosome Location
Chr.3: 49688753-49721529 on Build GRCh38
Transcripts
NM_198722.2 NM_022064.4 NR_135218.1 XM_006713285.1 XM_011533995.1 XM_017007018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6599260 c.898T>C p.C300R Chr.3: 49718568-49718568 on Build GRCh38
6599259 c.1039G>A p.A347T Chr.3: 49718427-49718427 on Build GRCh38
1182678 c.948G>T p.Q316H Chr.3: 49718518-49718518 on Build GRCh38
3427713 c.942G>A p.P314P Chr.3: 49718524-49718524 on Build GRCh38
1309281 c.888G>A p.L296L Chr.3: 49718578-49718578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018486205 Chr.3: 49718492-49718492 on Build GRCh38
rs772464782 Chr.3: 49718393-49718393 on Build GRCh38
rs758352733 Chr.3: 49718365-49718365 on Build GRCh38
rs779856258 Chr.3: 49718366-49718366 on Build GRCh38
rs867957408 Chr.3: 49718468-49718468 on Build GRCh38
rs1012519663 Chr.3: 49718349-49718349 on Build GRCh38
rs775062014 Chr.3: 49718336-49718336 on Build GRCh38
rs777353932 Chr.3: 49718466-49718466 on Build GRCh38
rs776880189 Chr.3: 49718412-49718412 on Build GRCh38
rs780494082 Chr.3: 49718477-49718477 on Build GRCh38
rs761947662 Chr.3: 49718356-49718356 on Build GRCh38
rs762999839 Chr.3: 49718520-49718520 on Build GRCh38
rs573198411 Chr.3: 49718449-49718449 on Build GRCh38
rs776138776 Chr.3: 49718330-49718330 on Build GRCh38
rs1030585913 Chr.3: 49718524-49718524 on Build GRCh38
rs575362033 Chr.3: 49718369-49718369 on Build GRCh38
rs1051795677 Chr.3: 49718415-49718415 on Build GRCh38
rs779537959 Chr.3: 49718382-49718382 on Build GRCh38
rs376736320 Chr.3: 49718441-49718441 on Build GRCh38
rs752548201 Chr.3: 49718540-49718540 on Build GRCh38
rs780762034 Chr.3: 49718371-49718371 on Build GRCh38
rs761004181 Chr.3: 49718398-49718398 on Build GRCh38
rs769465616 Chr.3: 49718374-49718374 on Build GRCh38
rs755772643 Chr.3: 49718552-49718552 on Build GRCh38
rs1041997599 Chr.3: 49718395-49718395 on Build GRCh38
rs774191576 Chr.3: 49718531-49718531 on Build GRCh38
rs747670903 Chr.3: 49718331-49718331 on Build GRCh38
rs368808320 Chr.3: 49718471-49718471 on Build GRCh38
rs753941544 Chr.3: 49718354-49718354 on Build GRCh38
rs903154052 Chr.3: 49718404-49718404 on Build GRCh38
rs755865197 Chr.3: 49718450-49718450 on Build GRCh38
rs766332423 Chr.3: 49718529-49718529 on Build GRCh38
rs747369013 Chr.3: 49718485-49718485 on Build GRCh38
rs767878062 Chr.3: 49718346-49718346 on Build GRCh38
rs889194560 Chr.3: 49718423-49718423 on Build GRCh38
rs868789247 Chr.3: 49718544-49718544 on Build GRCh38
rs750411107 Chr.3: 49718364-49718364 on Build GRCh38
rs748825526 Chr.3: 49718469-49718469 on Build GRCh38
rs537662999 Chr.3: 49718350-49718350 on Build GRCh38
rs765250301 Chr.3: 49718425-49718425 on Build GRCh38
rs374538101 Chr.3: 49718332-49718332 on Build GRCh38
rs199566932 Chr.3: 49718394-49718394 on Build GRCh38
rs760888415 Chr.3: 49718351-49718351 on Build GRCh38
rs768918710 Chr.3: 49718407-49718407 on Build GRCh38
rs981834081 Chr.3: 49718498-49718498 on Build GRCh38
rs780864622 Chr.3: 49718442-49718442 on Build GRCh38
rs778101475 Chr.3: 49718448-49718449 on Build GRCh38
rs62262720 Chr.3: 49718392-49718392 on Build GRCh38
rs34810303 Chr.3: 49718533-49718533 on Build GRCh38
rs768952020 Chr.3: 49718503-49718503 on Build GRCh38
rs557401374 Chr.3: 49718353-49718353 on Build GRCh38
rs759945468 Chr.3: 49718341-49718341 on Build GRCh38
rs761982260 Chr.3: 49718505-49718505 on Build GRCh38
rs766239635 Chr.3: 49718437-49718437 on Build GRCh38
rs138809316 Chr.3: 49718515-49718515 on Build GRCh38
rs765129698 Chr.3: 49718363-49718363 on Build GRCh38
rs750531309 Chr.3: 49718434-49718434 on Build GRCh38
rs962582792 Chr.3: 49718495-49718495 on Build GRCh38
rs747652352 Chr.3: 49718373-49718373 on Build GRCh38
rs749799985 Chr.3: 49718473-49718486 on Build GRCh38
rs751316970 Chr.3: 49718370-49718370 on Build GRCh38
rs762882013 Chr.3: 49718436-49718436 on Build GRCh38
rs767368457 Chr.3: 49718538-49718538 on Build GRCh38
rs539432112 Chr.3: 49718388-49718388 on Build GRCh38
rs761861468 Chr.3: 49718419-49718419 on Build GRCh38
rs199670077 Chr.3: 49718440-49718440 on Build GRCh38
rs771546841 Chr.3: 49718333-49718333 on Build GRCh38

View Full Product Details