Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 236264214-236264473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB18
Entrez Gene ID
401036
Gene Name
ankyrin repeat and SOCS box containing 18
Gene Aliases
ASB-18
UniGene
Hs.639909
Gene Chromosome Location
Chr.2: 236194872-236264345 on Build GRCh38
Transcripts
NM_212556.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3838976 c.92G>A p.R31K Chr.2: 236264254-236264254 on Build GRCh38
6450320 c.56G>C p.R19T Chr.2: 236264290-236264290 on Build GRCh38
6010729 c.23C>T p.P8L Chr.2: 236264323-236264323 on Build GRCh38
7319626 c.20T>A p.L7H Chr.2: 236264326-236264326 on Build GRCh38
335169 c.49G>T p.V17L Chr.2: 236264297-236264297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142164329 Chr.2: 236264394-236264394 on Build GRCh38
rs766976276 Chr.2: 236264282-236264282 on Build GRCh38
rs779743226 Chr.2: 236264337-236264337 on Build GRCh38
rs1016785990 Chr.2: 236264406-236264406 on Build GRCh38
rs765301252 Chr.2: 236264308-236264308 on Build GRCh38
rs753848625 Chr.2: 236264288-236264288 on Build GRCh38
rs746608036 Chr.2: 236264324-236264326 on Build GRCh38
rs764447024 Chr.2: 236264381-236264381 on Build GRCh38
rs758638427 Chr.2: 236264317-236264317 on Build GRCh38
rs370407156 Chr.2: 236264248-236264248 on Build GRCh38
rs751303042 Chr.2: 236264387-236264387 on Build GRCh38
rs188673979 Chr.2: 236264238-236264238 on Build GRCh38
rs199949640 Chr.2: 236264393-236264393 on Build GRCh38
rs200159870 Chr.2: 236264314-236264314 on Build GRCh38
rs886990228 Chr.2: 236264297-236264297 on Build GRCh38
rs895828352 Chr.2: 236264363-236264363 on Build GRCh38
rs146737753 Chr.2: 236264418-236264418 on Build GRCh38
rs763133723 Chr.2: 236264377-236264377 on Build GRCh38
rs754704683 Chr.2: 236264344-236264345 on Build GRCh38
rs756145947 Chr.2: 236264395-236264395 on Build GRCh38
rs372151498 Chr.2: 236264354-236264354 on Build GRCh38
rs910719893 Chr.2: 236264271-236264271 on Build GRCh38
rs777525281 Chr.2: 236264321-236264321 on Build GRCh38
rs758061404 Chr.2: 236264322-236264322 on Build GRCh38
rs779720714 Chr.2: 236264308-236264309 on Build GRCh38
rs768462060 Chr.2: 236264344-236264344 on Build GRCh38
rs990801581 Chr.2: 236264428-236264428 on Build GRCh38
rs996634859 Chr.2: 236264369-236264369 on Build GRCh38
rs772526346 Chr.2: 236264270-236264270 on Build GRCh38
rs567154088 Chr.2: 236264241-236264241 on Build GRCh38
rs748903039 Chr.2: 236264342-236264342 on Build GRCh38
rs375648650 Chr.2: 236264375-236264375 on Build GRCh38
rs773443450 Chr.2: 236264347-236264347 on Build GRCh38
rs201221674 Chr.2: 236264355-236264355 on Build GRCh38
rs759551732 Chr.2: 236264306-236264306 on Build GRCh38
rs771212696 Chr.2: 236264352-236264352 on Build GRCh38
rs780248286 Chr.2: 236264417-236264417 on Build GRCh38
rs942339623 Chr.2: 236264436-236264436 on Build GRCh38
rs762700857 Chr.2: 236264250-236264250 on Build GRCh38
rs747462961 Chr.2: 236264351-236264351 on Build GRCh38
rs769557775 Chr.2: 236264244-236264244 on Build GRCh38
rs769363208 Chr.2: 236264335-236264335 on Build GRCh38
rs1028054683 Chr.2: 236264370-236264370 on Build GRCh38
rs757142011 Chr.2: 236264324-236264324 on Build GRCh38
rs373891164 Chr.2: 236264279-236264279 on Build GRCh38
rs765248287 Chr.2: 236264366-236264366 on Build GRCh38
rs781251806 Chr.2: 236264334-236264334 on Build GRCh38

View Full Product Details