Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 43717618-43717891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD5
Entrez Gene ID
51099
Gene Name
abhydrolase domain containing 5
Gene Aliases
CDS, CGI58, IECN2, NCIE2
UniGene
Hs.19385
Gene Chromosome Location
Chr.3: 43690883-43722725 on Build GRCh38
Transcripts
NM_016006.4 XM_011533779.2 XM_011533780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7141432 c.839G>T p.R280L Chr.3: 43717736-43717736 on Build GRCh38
1044897 c.922A>G p.I308V Chr.3: 43717819-43717819 on Build GRCh38
6590900 c.819delA p.R275fs*18 Chr.3: 43717716-43717716 on Build GRCh38
1617608 c.829A>G p.M277V Chr.3: 43717726-43717726 on Build GRCh38
730816 c.890G>C p.R297P Chr.3: 43717787-43717787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773285310 Chr.3: 43717703-43717703 on Build GRCh38
rs774687594 Chr.3: 43717819-43717819 on Build GRCh38
rs368953991 Chr.3: 43717838-43717838 on Build GRCh38
rs537619304 Chr.3: 43717717-43717717 on Build GRCh38
rs746970260 Chr.3: 43717782-43717782 on Build GRCh38
rs763361774 Chr.3: 43717783-43717784 on Build GRCh38
rs750266230 Chr.3: 43717753-43717753 on Build GRCh38
rs28939078 Chr.3: 43717675-43717675 on Build GRCh38
rs775381851 Chr.3: 43717677-43717677 on Build GRCh38
rs758156503 Chr.3: 43717759-43717759 on Build GRCh38
rs866584094 Chr.3: 43717714-43717714 on Build GRCh38
rs574148007 Chr.3: 43717805-43717805 on Build GRCh38
rs377553984 Chr.3: 43717683-43717683 on Build GRCh38
rs140627711 Chr.3: 43717783-43717783 on Build GRCh38
rs773892644 Chr.3: 43717733-43717733 on Build GRCh38
rs756873141 Chr.3: 43717661-43717661 on Build GRCh38
rs759959911 Chr.3: 43717733-43717733 on Build GRCh38
rs764907762 Chr.3: 43717855-43717855 on Build GRCh38
rs987013194 Chr.3: 43717664-43717664 on Build GRCh38
rs746791416 Chr.3: 43717815-43717815 on Build GRCh38
rs748919068 Chr.3: 43717657-43717657 on Build GRCh38
rs778521688 Chr.3: 43717752-43717752 on Build GRCh38
rs775943260 Chr.3: 43717827-43717827 on Build GRCh38
rs763521549 Chr.3: 43717748-43717748 on Build GRCh38
rs763091024 Chr.3: 43717716-43717716 on Build GRCh38
rs745654978 Chr.3: 43717674-43717674 on Build GRCh38
rs761087968 Chr.3: 43717831-43717831 on Build GRCh38
rs768126868 Chr.3: 43717646-43717646 on Build GRCh38
rs753292128 Chr.3: 43717737-43717737 on Build GRCh38
rs980094867 Chr.3: 43717803-43717803 on Build GRCh38
rs894423675 Chr.3: 43717862-43717862 on Build GRCh38
rs908673218 Chr.3: 43717670-43717670 on Build GRCh38
rs764653478 Chr.3: 43717832-43717832 on Build GRCh38
rs771108440 Chr.3: 43717809-43717809 on Build GRCh38
rs774914927 Chr.3: 43717732-43717732 on Build GRCh38
rs369544279 Chr.3: 43717691-43717691 on Build GRCh38
rs892335181 Chr.3: 43717736-43717736 on Build GRCh38
rs886058488 Chr.3: 43717803-43717803 on Build GRCh38
rs772591700 Chr.3: 43717826-43717826 on Build GRCh38
rs777783278 Chr.3: 43717791-43717791 on Build GRCh38
rs554355585 Chr.3: 43717738-43717738 on Build GRCh38
rs752307149 Chr.3: 43717647-43717647 on Build GRCh38
rs370301465 Chr.3: 43717735-43717735 on Build GRCh38
rs542974582 Chr.3: 43717749-43717749 on Build GRCh38
rs1030871913 Chr.3: 43717695-43717695 on Build GRCh38
rs147129794 Chr.3: 43717776-43717776 on Build GRCh38
rs777300743 Chr.3: 43717648-43717648 on Build GRCh38
rs374342059 Chr.3: 43717663-43717663 on Build GRCh38
rs930353467 Chr.3: 43717784-43717784 on Build GRCh38

View Full Product Details