Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3910678-3910933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATCAY
Entrez Gene ID
85300
Gene Name
ATCAY, caytaxin
Gene Aliases
BNIP-H, CLAC
UniGene
Hs.418055
Gene Chromosome Location
Chr.19: 3880620-3928082 on Build GRCh38
Transcripts
NM_033064.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5811279 c.822G>T p.S274S Chr.19: 3910845-3910845 on Build GRCh38
3533546 c.861C>T p.F287F Chr.19: 3910884-3910884 on Build GRCh38
1392697 c.780-1G>T p.? Chr.19: 3910802-3910802 on Build GRCh38
1392696 c.780-1G>T p.? Chr.19: 3910802-3910802 on Build GRCh38
996102 c.785G>A p.R262Q Chr.19: 3910808-3910808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756637199 Chr.19: 3910877-3910877 on Build GRCh38
rs746110322 Chr.19: 3910897-3910897 on Build GRCh38
rs761815055 Chr.19: 3910790-3910790 on Build GRCh38
rs764564106 Chr.19: 3910878-3910878 on Build GRCh38
rs1014676893 Chr.19: 3910842-3910842 on Build GRCh38
rs974592593 Chr.19: 3910811-3910811 on Build GRCh38
rs202054448 Chr.19: 3910906-3910906 on Build GRCh38
rs753930037 Chr.19: 3910882-3910882 on Build GRCh38
rs753075939 Chr.19: 3910874-3910874 on Build GRCh38
rs370449188 Chr.19: 3910761-3910761 on Build GRCh38
rs768469974 Chr.19: 3910787-3910787 on Build GRCh38
rs375049279 Chr.19: 3910899-3910899 on Build GRCh38
rs1043599636 Chr.19: 3910718-3910718 on Build GRCh38
rs768004227 Chr.19: 3910863-3910863 on Build GRCh38
rs376658701 Chr.19: 3910855-3910855 on Build GRCh38
rs1057457018 Chr.19: 3910903-3910903 on Build GRCh38
rs1013447061 Chr.19: 3910912-3910912 on Build GRCh38
rs116660680 Chr.19: 3910895-3910895 on Build GRCh38
rs75191480 Chr.19: 3910711-3910711 on Build GRCh38
rs920462404 Chr.19: 3910815-3910815 on Build GRCh38
rs761281770 Chr.19: 3910887-3910887 on Build GRCh38
rs1031294020 Chr.19: 3910707-3910707 on Build GRCh38
rs372306459 Chr.19: 3910845-3910845 on Build GRCh38
rs61746441 Chr.19: 3910833-3910833 on Build GRCh38
rs372724058 Chr.19: 3910913-3910913 on Build GRCh38
rs776502708 Chr.19: 3910789-3910789 on Build GRCh38
rs987590673 Chr.19: 3910708-3910708 on Build GRCh38
rs554936677 Chr.19: 3910731-3910731 on Build GRCh38
rs575418065 Chr.19: 3910712-3910712 on Build GRCh38
rs767922336 Chr.19: 3910717-3910717 on Build GRCh38
rs766102010 Chr.19: 3910825-3910825 on Build GRCh38
rs373667210 Chr.19: 3910762-3910762 on Build GRCh38
rs769688062 Chr.19: 3910795-3910795 on Build GRCh38
rs762422605 Chr.19: 3910821-3910821 on Build GRCh38
rs544396501 Chr.19: 3910730-3910730 on Build GRCh38
rs140799527 Chr.19: 3910896-3910896 on Build GRCh38
rs772767948 Chr.19: 3910798-3910798 on Build GRCh38

View Full Product Details