Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 65072312-65072539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP68
Entrez Gene ID
23177
Gene Name
centrosomal protein 68
Gene Aliases
KIAA0582
UniGene
Hs.709257
Gene Chromosome Location
Chr.2: 65056361-65087008 on Build GRCh38
Transcripts
NM_001319100.1 NM_001319101.1 NM_015147.2 NR_134966.1 XM_005264219.3 XM_017003633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
345347 c.1374G>C p.Q458H Chr.2: 65072470-65072470 on Build GRCh38
1200834 c.1321C>T p.R441W Chr.2: 65072417-65072417 on Build GRCh38
3582590 c.175T>C p.S59P Chr.2: 65072435-65072435 on Build GRCh38
319417 c.1438G>A p.D480N Chr.2: 65072534-65072534 on Build GRCh38
5862715 c.118G>A p.G40S Chr.2: 65072378-65072378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921894381 Chr.2: 65072478-65072478 on Build GRCh38
rs769316304 Chr.2: 65072404-65072404 on Build GRCh38
rs577037216 Chr.2: 65072366-65072366 on Build GRCh38
rs768797246 Chr.2: 65072336-65072336 on Build GRCh38
rs764888119 Chr.2: 65072436-65072436 on Build GRCh38
rs752630437 Chr.2: 65072350-65072350 on Build GRCh38
rs150135845 Chr.2: 65072514-65072514 on Build GRCh38
rs149395099 Chr.2: 65072427-65072427 on Build GRCh38
rs140998700 Chr.2: 65072337-65072337 on Build GRCh38
rs746213881 Chr.2: 65072394-65072394 on Build GRCh38
rs141435576 Chr.2: 65072333-65072333 on Build GRCh38
rs141670878 Chr.2: 65072375-65072375 on Build GRCh38
rs755795918 Chr.2: 65072481-65072481 on Build GRCh38
rs777917866 Chr.2: 65072515-65072515 on Build GRCh38
rs980009597 Chr.2: 65072406-65072406 on Build GRCh38
rs147665059 Chr.2: 65072477-65072477 on Build GRCh38
rs35089924 Chr.2: 65072513-65072513 on Build GRCh38
rs764044528 Chr.2: 65072460-65072460 on Build GRCh38
rs143142350 Chr.2: 65072359-65072359 on Build GRCh38
rs759285980 Chr.2: 65072347-65072347 on Build GRCh38
rs149863693 Chr.2: 65072437-65072437 on Build GRCh38
rs1018760416 Chr.2: 65072469-65072469 on Build GRCh38
rs754249961 Chr.2: 65072486-65072486 on Build GRCh38
rs935592823 Chr.2: 65072499-65072499 on Build GRCh38
rs1014021266 Chr.2: 65072414-65072414 on Build GRCh38
rs774522142 Chr.2: 65072417-65072417 on Build GRCh38
rs767871813 Chr.2: 65072345-65072345 on Build GRCh38
rs772389739 Chr.2: 65072424-65072424 on Build GRCh38
rs762085923 Chr.2: 65072341-65072341 on Build GRCh38
rs145806974 Chr.2: 65072490-65072490 on Build GRCh38
rs752444166 Chr.2: 65072416-65072417 on Build GRCh38
rs971023170 Chr.2: 65072367-65072367 on Build GRCh38
rs752463713 Chr.2: 65072358-65072358 on Build GRCh38
rs150680996 Chr.2: 65072418-65072418 on Build GRCh38
rs895549364 Chr.2: 65072409-65072409 on Build GRCh38
rs183629174 Chr.2: 65072398-65072398 on Build GRCh38
rs765212445 Chr.2: 65072348-65072348 on Build GRCh38
rs780480482 Chr.2: 65072400-65072400 on Build GRCh38
rs761354792 Chr.2: 65072474-65072474 on Build GRCh38
rs780873466 Chr.2: 65072393-65072393 on Build GRCh38
rs751048637 Chr.2: 65072464-65072464 on Build GRCh38
rs35694840 Chr.2: 65072480-65072480 on Build GRCh38
rs773369946 Chr.2: 65072426-65072426 on Build GRCh38
rs542392624 Chr.2: 65072484-65072484 on Build GRCh38
rs762840356 Chr.2: 65072440-65072440 on Build GRCh38
rs780240720 Chr.2: 65072483-65072483 on Build GRCh38
rs779601426 Chr.2: 65072498-65072498 on Build GRCh38
rs137906764 Chr.2: 65072383-65072383 on Build GRCh38
rs199831101 Chr.2: 65072420-65072420 on Build GRCh38
rs749690692 Chr.2: 65072402-65072402 on Build GRCh38

View Full Product Details