Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 80155730-80155997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA11
Entrez Gene ID
311
Gene Name
annexin A11
Gene Aliases
ANX11, CAP50
UniGene
Hs.530291
Gene Chromosome Location
Chr.10: 80153953-80205677 on Build GRCh38
Transcripts
NM_001157.2 NM_001278407.1 NM_001278408.1 NM_001278409.1 NM_145868.1 NM_145869.1 XM_005269741.4 XM_011539736.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4016255 c.1505G>A p.G502D Chr.10: 80155866-80155866 on Build GRCh38
1966624 c.1498A>T p.I500F Chr.10: 80155873-80155873 on Build GRCh38
236990 c.1459-5G>A p.? Chr.10: 80155917-80155917 on Build GRCh38
3931138 c.1508G>A p.G503D Chr.10: 80155863-80155863 on Build GRCh38
6448308 c.1480C>T p.R494W Chr.10: 80155891-80155891 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779661783 Chr.10: 80155829-80155829 on Build GRCh38
rs747039260 Chr.10: 80155891-80155891 on Build GRCh38
rs776283120 Chr.10: 80155843-80155843 on Build GRCh38
rs866169122 Chr.10: 80155756-80155756 on Build GRCh38
rs765906318 Chr.10: 80155933-80155933 on Build GRCh38
rs757971318 Chr.10: 80155827-80155827 on Build GRCh38
rs748998938 Chr.10: 80155964-80155964 on Build GRCh38
rs751091287 Chr.10: 80155940-80155940 on Build GRCh38
rs190607138 Chr.10: 80155864-80155864 on Build GRCh38
rs951314626 Chr.10: 80155915-80155915 on Build GRCh38
rs200104175 Chr.10: 80155849-80155849 on Build GRCh38
rs765688945 Chr.10: 80155906-80155906 on Build GRCh38
rs867251586 Chr.10: 80155908-80155908 on Build GRCh38
rs1007654038 Chr.10: 80155754-80155754 on Build GRCh38
rs200609038 Chr.10: 80155878-80155878 on Build GRCh38
rs369415003 Chr.10: 80155834-80155834 on Build GRCh38
rs777207980 Chr.10: 80155900-80155900 on Build GRCh38
rs376530605 Chr.10: 80155814-80155814 on Build GRCh38
rs143332492 Chr.10: 80155818-80155818 on Build GRCh38
rs540903210 Chr.10: 80155819-80155819 on Build GRCh38
rs928322155 Chr.10: 80155939-80155939 on Build GRCh38
rs772154973 Chr.10: 80155871-80155871 on Build GRCh38
rs867436516 Chr.10: 80155949-80155949 on Build GRCh38
rs561680336 Chr.10: 80155753-80155753 on Build GRCh38
rs962558601 Chr.10: 80155785-80155785 on Build GRCh38
rs201989810 Chr.10: 80155825-80155825 on Build GRCh38
rs1802934 Chr.10: 80155772-80155772 on Build GRCh38
rs773424974 Chr.10: 80155920-80155920 on Build GRCh38
rs764508534 Chr.10: 80155873-80155875 on Build GRCh38
rs113509756 Chr.10: 80155969-80155969 on Build GRCh38
rs781057466 Chr.10: 80155840-80155840 on Build GRCh38
rs762218493 Chr.10: 80155905-80155905 on Build GRCh38
rs1018518861 Chr.10: 80155781-80155781 on Build GRCh38
rs375921939 Chr.10: 80155805-80155805 on Build GRCh38
rs755985045 Chr.10: 80155954-80155954 on Build GRCh38
rs78664479 Chr.10: 80155948-80155948 on Build GRCh38
rs958492281 Chr.10: 80155972-80155972 on Build GRCh38
rs762214872 Chr.10: 80155823-80155823 on Build GRCh38
rs762705669 Chr.10: 80155930-80155930 on Build GRCh38
rs769304031 Chr.10: 80155898-80155898 on Build GRCh38
rs77908721 Chr.10: 80155910-80155910 on Build GRCh38
rs752690669 Chr.10: 80155953-80155953 on Build GRCh38
rs746152448 Chr.10: 80155866-80155866 on Build GRCh38
rs777665525 Chr.10: 80155963-80155963 on Build GRCh38
rs767010177 Chr.10: 80155951-80155951 on Build GRCh38

View Full Product Details