Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6730624-6730867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT5
Entrez Gene ID
55326
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 5
Gene Aliases
1AGPAT5, LPAATE
UniGene
Hs.624002
Gene Chromosome Location
Chr.8: 6708357-6761503 on Build GRCh38
Transcripts
NM_018361.3 XM_011534751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5979492 c.316G>T p.A106S Chr.8: 6730737-6730737 on Build GRCh38
4441287 c.323G>T p.R108M Chr.8: 6730744-6730744 on Build GRCh38
3774336 c.384G>T p.L128F Chr.8: 6730805-6730805 on Build GRCh38
3900971 c.346C>T p.R116C Chr.8: 6730767-6730767 on Build GRCh38
1624122 c.310A>G p.I104V Chr.8: 6730731-6730731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767417802 Chr.8: 6730740-6730740 on Build GRCh38
rs757728859 Chr.8: 6730705-6730705 on Build GRCh38
rs145228795 Chr.8: 6730737-6730737 on Build GRCh38
rs138829950 Chr.8: 6730753-6730753 on Build GRCh38
rs776513670 Chr.8: 6730820-6730820 on Build GRCh38
rs754720986 Chr.8: 6730694-6730694 on Build GRCh38
rs770132539 Chr.8: 6730725-6730725 on Build GRCh38
rs769937996 Chr.8: 6730668-6730668 on Build GRCh38
rs370412609 Chr.8: 6730671-6730671 on Build GRCh38
rs1019580232 Chr.8: 6730649-6730649 on Build GRCh38
rs139817148 Chr.8: 6730734-6730734 on Build GRCh38
rs960613263 Chr.8: 6730700-6730700 on Build GRCh38
rs768465049 Chr.8: 6730679-6730680 on Build GRCh38
rs760748971 Chr.8: 6730749-6730749 on Build GRCh38
rs965561403 Chr.8: 6730651-6730651 on Build GRCh38
rs749479144 Chr.8: 6730731-6730731 on Build GRCh38
rs768865359 Chr.8: 6730771-6730771 on Build GRCh38
rs766341895 Chr.8: 6730687-6730687 on Build GRCh38
rs773473423 Chr.8: 6730792-6730792 on Build GRCh38
rs779002902 Chr.8: 6730773-6730773 on Build GRCh38
rs570143152 Chr.8: 6730680-6730680 on Build GRCh38
rs201844845 Chr.8: 6730674-6730674 on Build GRCh38
rs375065662 Chr.8: 6730660-6730660 on Build GRCh38
rs774332640 Chr.8: 6730675-6730675 on Build GRCh38
rs756652472 Chr.8: 6730767-6730767 on Build GRCh38
rs748466250 Chr.8: 6730781-6730781 on Build GRCh38
rs752279124 Chr.8: 6730830-6730830 on Build GRCh38
rs367702364 Chr.8: 6730772-6730772 on Build GRCh38
rs533474603 Chr.8: 6730659-6730659 on Build GRCh38
rs772028188 Chr.8: 6730788-6730788 on Build GRCh38
rs200898253 Chr.8: 6730684-6730684 on Build GRCh38
rs768790339 Chr.8: 6730733-6730733 on Build GRCh38
rs200945982 Chr.8: 6730656-6730656 on Build GRCh38
rs753524722 Chr.8: 6730692-6730692 on Build GRCh38
rs762717911 Chr.8: 6730831-6730831 on Build GRCh38
rs746441349 Chr.8: 6730708-6730708 on Build GRCh38
rs374362603 Chr.8: 6730685-6730685 on Build GRCh38
rs760652597 Chr.8: 6730797-6730797 on Build GRCh38
rs376636029 Chr.8: 6730757-6730757 on Build GRCh38
rs746248999 Chr.8: 6730664-6730664 on Build GRCh38
rs201930945 Chr.8: 6730825-6730825 on Build GRCh38
rs552205808 Chr.8: 6730761-6730761 on Build GRCh38
rs777010150 Chr.8: 6730662-6730662 on Build GRCh38
rs773922038 Chr.8: 6730723-6730733 on Build GRCh38
rs368467009 Chr.8: 6730703-6730703 on Build GRCh38
rs765146947 Chr.8: 6730822-6730822 on Build GRCh38
rs1048955179 Chr.8: 6730759-6730759 on Build GRCh38
rs115348738 Chr.8: 6730802-6730802 on Build GRCh38
rs908568858 Chr.8: 6730707-6730707 on Build GRCh38
rs778449158 Chr.8: 6730701-6730701 on Build GRCh38
rs372429394 Chr.8: 6730756-6730756 on Build GRCh38
rs753719874 Chr.8: 6730754-6730754 on Build GRCh38
rs985466697 Chr.8: 6730647-6730647 on Build GRCh38
rs990286620 Chr.8: 6730676-6730676 on Build GRCh38
rs141780106 Chr.8: 6730763-6730763 on Build GRCh38
rs755973461 Chr.8: 6730686-6730686 on Build GRCh38
rs749675241 Chr.8: 6730769-6730769 on Build GRCh38
rs773275147 Chr.8: 6730748-6730748 on Build GRCh38

View Full Product Details