Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134144178-134144451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAM3
Entrez Gene ID
83700
Gene Name
junctional adhesion molecule 3
Gene Aliases
JAM-2, JAM-3, JAM-C, JAMC
UniGene
Hs.150718
Gene Chromosome Location
Chr.11: 134068925-134151757 on Build GRCh38
Transcripts
NM_001205329.1 NM_032801.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7186663 c.392-3T>G p.? Chr.11: 134144238-134144238 on Build GRCh38
1255150 c.392-2_392delAGG p.? Chr.11: 134144239-134144241 on Build GRCh38
7139699 c.454C>T p.R152W Chr.11: 134144303-134144303 on Build GRCh38
687177 c.433C>T p.L145L Chr.11: 134144282-134144282 on Build GRCh38
5483532 c.499G>A p.D167N Chr.11: 134144348-134144348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376093579 Chr.11: 134144221-134144221 on Build GRCh38
rs774439815 Chr.11: 134144228-134144228 on Build GRCh38
rs536759201 Chr.11: 134144335-134144335 on Build GRCh38
rs746028100 Chr.11: 134144408-134144408 on Build GRCh38
rs189148616 Chr.11: 134144250-134144250 on Build GRCh38
rs776039938 Chr.11: 134144237-134144237 on Build GRCh38
rs746796075 Chr.11: 134144217-134144217 on Build GRCh38
rs35689917 Chr.11: 134144356-134144356 on Build GRCh38
rs781039978 Chr.11: 134144372-134144372 on Build GRCh38
rs763314592 Chr.11: 134144222-134144224 on Build GRCh38
rs377687676 Chr.11: 134144225-134144225 on Build GRCh38
rs762709121 Chr.11: 134144343-134144343 on Build GRCh38
rs762126235 Chr.11: 134144336-134144336 on Build GRCh38
rs749919619 Chr.11: 134144262-134144262 on Build GRCh38
rs750102309 Chr.11: 134144342-134144342 on Build GRCh38
rs977837404 Chr.11: 134144352-134144352 on Build GRCh38
rs267602787 Chr.11: 134144256-134144256 on Build GRCh38
rs200509708 Chr.11: 134144385-134144385 on Build GRCh38
rs745951708 Chr.11: 134144303-134144303 on Build GRCh38
rs758048688 Chr.11: 134144267-134144267 on Build GRCh38
rs754969540 Chr.11: 134144272-134144272 on Build GRCh38
rs926193098 Chr.11: 134144355-134144355 on Build GRCh38
rs140641472 Chr.11: 134144251-134144251 on Build GRCh38
rs771087930 Chr.11: 134144226-134144226 on Build GRCh38
rs747916870 Chr.11: 134144279-134144279 on Build GRCh38
rs148009046 Chr.11: 134144269-134144269 on Build GRCh38
rs752502218 Chr.11: 134144377-134144377 on Build GRCh38
rs370223986 Chr.11: 134144253-134144253 on Build GRCh38
rs780183089 Chr.11: 134144211-134144211 on Build GRCh38
rs761134261 Chr.11: 134144324-134144324 on Build GRCh38
rs1026347341 Chr.11: 134144247-134144247 on Build GRCh38
rs975702687 Chr.11: 134144411-134144411 on Build GRCh38
rs371389064 Chr.11: 134144201-134144201 on Build GRCh38
rs1016830203 Chr.11: 134144219-134144219 on Build GRCh38
rs764938157 Chr.11: 134144339-134144339 on Build GRCh38
rs397515439 Chr.11: 134144330-134144330 on Build GRCh38
rs750770795 Chr.11: 134144212-134144212 on Build GRCh38
rs771159725 Chr.11: 134144290-134144290 on Build GRCh38
rs767979226 Chr.11: 134144235-134144235 on Build GRCh38
rs1042196839 Chr.11: 134144383-134144383 on Build GRCh38
rs769175735 Chr.11: 134144325-134144325 on Build GRCh38
rs779180039 Chr.11: 134144386-134144386 on Build GRCh38
rs780997837 Chr.11: 134144274-134144274 on Build GRCh38
rs775555456 Chr.11: 134144304-134144304 on Build GRCh38
rs541627406 Chr.11: 134144301-134144301 on Build GRCh38
rs779041471 Chr.11: 134144292-134144292 on Build GRCh38
rs759580323 Chr.11: 134144230-134144230 on Build GRCh38
rs374174597 Chr.11: 134144351-134144351 on Build GRCh38
rs1033470182 Chr.11: 134144245-134144245 on Build GRCh38
rs555033192 Chr.11: 134144360-134144360 on Build GRCh38
rs776580488 Chr.11: 134144223-134144223 on Build GRCh38

View Full Product Details