Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 224157319-224157592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO28
Entrez Gene ID
23219
Gene Name
F-box protein 28
Gene Aliases
CENP-30, Fbx28
UniGene
Hs.64691
Gene Chromosome Location
Chr.1: 224114087-224162047 on Build GRCh38
Transcripts
NM_001136115.2 NM_015176.3 NR_049764.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3484170 c.865G>T p.V289L Chr.1: 224157504-224157504 on Build GRCh38
1178044 c.894C>T p.D298D Chr.1: 224157533-224157533 on Build GRCh38
5810487 c.807A>T p.K269N Chr.1: 224157446-224157446 on Build GRCh38
6662155 c.947G>A p.R316Q Chr.1: 224157586-224157586 on Build GRCh38
679403 c.713T>G p.V238G Chr.1: 224157352-224157352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758418791 Chr.1: 224157560-224157560 on Build GRCh38
rs760205388 Chr.1: 224157403-224157403 on Build GRCh38
rs777854711 Chr.1: 224157566-224157566 on Build GRCh38
rs754071147 Chr.1: 224157352-224157352 on Build GRCh38
rs769466378 Chr.1: 224157445-224157445 on Build GRCh38
rs767030831 Chr.1: 224157495-224157495 on Build GRCh38
rs755352864 Chr.1: 224157499-224157499 on Build GRCh38
rs562189685 Chr.1: 224157371-224157371 on Build GRCh38
rs544337297 Chr.1: 224157416-224157416 on Build GRCh38
rs773804810 Chr.1: 224157474-224157474 on Build GRCh38
rs189250147 Chr.1: 224157365-224157365 on Build GRCh38
rs201209850 Chr.1: 224157362-224157362 on Build GRCh38
rs754206716 Chr.1: 224157496-224157496 on Build GRCh38
rs752869868 Chr.1: 224157517-224157517 on Build GRCh38
rs1023471990 Chr.1: 224157471-224157471 on Build GRCh38
rs866661926 Chr.1: 224157534-224157534 on Build GRCh38
rs763710165 Chr.1: 224157466-224157466 on Build GRCh38
rs762458333 Chr.1: 224157462-224157462 on Build GRCh38
rs770666477 Chr.1: 224157408-224157408 on Build GRCh38
rs146604018 Chr.1: 224157461-224157461 on Build GRCh38
rs745691947 Chr.1: 224157419-224157419 on Build GRCh38
rs772583199 Chr.1: 224157468-224157468 on Build GRCh38
rs771426966 Chr.1: 224157361-224157361 on Build GRCh38
rs761294837 Chr.1: 224157478-224157478 on Build GRCh38
rs765488486 Chr.1: 224157509-224157509 on Build GRCh38
rs973258022 Chr.1: 224157470-224157470 on Build GRCh38
rs115866173 Chr.1: 224157407-224157407 on Build GRCh38

View Full Product Details