Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156596320-156596547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH4
Entrez Gene ID
54865
Gene Name
G-patch domain containing 4
Gene Aliases
GPATC4
UniGene
Hs.193832
Gene Chromosome Location
Chr.1: 156594308-156601496 on Build GRCh38
Transcripts
NM_015590.3 NM_182679.2 XM_005245287.3 XM_017000319.1

Gene Symbol
NAXE
Entrez Gene ID
128240
Gene Name
NAD(P)HX epimerase
Gene Aliases
AIBP, APOA1BP, YJEFN1
UniGene
Hs.528320
Gene Chromosome Location
Chr.1: 156591762-156599818 on Build GRCh38
Transcripts
NM_015590.3 NM_182679.2 XM_005245287.3 XM_017000319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3475319 c.349-1G>A p.? Chr.1: 156596479-156596479 on Build GRCh38
3705085 c.334-1G>C p.? Chr.1: 156596479-156596479 on Build GRCh38
4687736 c.378G>A p.K126K Chr.1: 156596449-156596449 on Build GRCh38
4788914 c.371A>C p.K124T Chr.1: 156596441-156596441 on Build GRCh38
3475320 c.334-1G>A p.? Chr.1: 156596479-156596479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770846200 Chr.1: 156596492-156596492 on Build GRCh38
rs773034872 Chr.1: 156596405-156596405 on Build GRCh38
rs776496861 Chr.1: 156596394-156596394 on Build GRCh38
rs775855443 Chr.1: 156596421-156596421 on Build GRCh38
rs750752156 Chr.1: 156596488-156596488 on Build GRCh38
rs754687136 Chr.1: 156596508-156596508 on Build GRCh38
rs757749375 Chr.1: 156596469-156596469 on Build GRCh38
rs779615333 Chr.1: 156596497-156596497 on Build GRCh38
rs34776980 Chr.1: 156596460-156596460 on Build GRCh38
rs768163432 Chr.1: 156596369-156596369 on Build GRCh38
rs1063288 Chr.1: 156596419-156596419 on Build GRCh38
rs777697309 Chr.1: 156596413-156596415 on Build GRCh38
rs751230320 Chr.1: 156596500-156596500 on Build GRCh38
rs74974163 Chr.1: 156596494-156596494 on Build GRCh38
rs1063287 Chr.1: 156596422-156596422 on Build GRCh38
rs137974014 Chr.1: 156596403-156596403 on Build GRCh38
rs936331431 Chr.1: 156596391-156596391 on Build GRCh38
rs141804813 Chr.1: 156596416-156596416 on Build GRCh38
rs754036498 Chr.1: 156596437-156596437 on Build GRCh38
rs147014962 Chr.1: 156596404-156596404 on Build GRCh38
rs762675705 Chr.1: 156596415-156596415 on Build GRCh38
rs746482378 Chr.1: 156596364-156596364 on Build GRCh38
rs749152834 Chr.1: 156596434-156596440 on Build GRCh38
rs905019550 Chr.1: 156596436-156596436 on Build GRCh38
rs765714345 Chr.1: 156596485-156596485 on Build GRCh38
rs760779293 Chr.1: 156596426-156596426 on Build GRCh38
rs200989966 Chr.1: 156596427-156596427 on Build GRCh38
rs1130058 Chr.1: 156596397-156596397 on Build GRCh38
rs758330231 Chr.1: 156596366-156596366 on Build GRCh38
rs777750007 Chr.1: 156596408-156596408 on Build GRCh38
rs779703992 Chr.1: 156596348-156596348 on Build GRCh38
rs758049447 Chr.1: 156596345-156596345 on Build GRCh38

View Full Product Details