Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27082799-27083056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMILIN1
Entrez Gene ID
11117
Gene Name
elastin microfibril interfacer 1
Gene Aliases
EMI, EMILIN, gp115
UniGene
Hs.63348
Gene Chromosome Location
Chr.2: 27078567-27086403 on Build GRCh38
Transcripts
NM_007046.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
368731 c.1347G>T p.L449F Chr.2: 27082918-27082918 on Build GRCh38
1407361 c.1467G>T p.Q489H Chr.2: 27083038-27083038 on Build GRCh38
2913097 c.1434C>T p.C478C Chr.2: 27083005-27083005 on Build GRCh38
5993833 c.1395T>C p.D465D Chr.2: 27082966-27082966 on Build GRCh38
4312396 c.1457C>A p.P486H Chr.2: 27083028-27083028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775872153 Chr.2: 27082937-27082938 on Build GRCh38
rs539737004 Chr.2: 27082924-27082924 on Build GRCh38
rs982822559 Chr.2: 27082822-27082822 on Build GRCh38
rs763633527 Chr.2: 27082917-27082918 on Build GRCh38
rs780817373 Chr.2: 27082976-27082976 on Build GRCh38
rs952149419 Chr.2: 27082869-27082869 on Build GRCh38
rs770081198 Chr.2: 27082882-27082882 on Build GRCh38
rs778862004 Chr.2: 27082866-27082866 on Build GRCh38
rs558070418 Chr.2: 27082925-27082925 on Build GRCh38
rs567811350 Chr.2: 27082827-27082827 on Build GRCh38
rs761144034 Chr.2: 27083007-27083007 on Build GRCh38
rs766813380 Chr.2: 27082899-27082899 on Build GRCh38
rs761454534 Chr.2: 27082951-27082951 on Build GRCh38
rs765615373 Chr.2: 27082856-27082856 on Build GRCh38
rs570508845 Chr.2: 27082919-27082919 on Build GRCh38
rs775866025 Chr.2: 27082947-27082947 on Build GRCh38
rs749590273 Chr.2: 27082989-27082989 on Build GRCh38
rs767934651 Chr.2: 27082916-27082916 on Build GRCh38
rs752386125 Chr.2: 27082833-27082833 on Build GRCh38
rs749848971 Chr.2: 27082954-27082954 on Build GRCh38
rs755321231 Chr.2: 27082821-27082821 on Build GRCh38
rs774287900 Chr.2: 27082939-27082939 on Build GRCh38
rs759927653 Chr.2: 27082907-27082907 on Build GRCh38
rs535162284 Chr.2: 27082865-27082865 on Build GRCh38
rs769135390 Chr.2: 27083031-27083031 on Build GRCh38
rs779056315 Chr.2: 27082994-27082994 on Build GRCh38
rs560031379 Chr.2: 27082953-27082953 on Build GRCh38
rs371009871 Chr.2: 27082959-27082959 on Build GRCh38
rs763045645 Chr.2: 27082898-27082898 on Build GRCh38
rs772502689 Chr.2: 27082943-27082943 on Build GRCh38
rs374226054 Chr.2: 27083005-27083005 on Build GRCh38
rs368076283 Chr.2: 27082818-27082818 on Build GRCh38
rs757174626 Chr.2: 27082861-27082861 on Build GRCh38
rs748766902 Chr.2: 27082849-27082849 on Build GRCh38
rs577341126 Chr.2: 27082942-27082942 on Build GRCh38
rs775777386 Chr.2: 27082923-27082923 on Build GRCh38
rs774971088 Chr.2: 27082905-27082905 on Build GRCh38
rs745910345 Chr.2: 27083002-27083002 on Build GRCh38
rs751896619 Chr.2: 27082819-27082819 on Build GRCh38
rs772037121 Chr.2: 27082875-27082875 on Build GRCh38
rs773426833 Chr.2: 27082889-27082889 on Build GRCh38
rs771044705 Chr.2: 27082991-27082991 on Build GRCh38
rs764421233 Chr.2: 27082952-27082952 on Build GRCh38
rs760050287 Chr.2: 27082842-27082844 on Build GRCh38
rs557123421 Chr.2: 27082918-27082918 on Build GRCh38
rs371224998 Chr.2: 27082920-27082920 on Build GRCh38
rs777280595 Chr.2: 27082839-27082839 on Build GRCh38
rs780004975 Chr.2: 27082922-27082922 on Build GRCh38
rs760979319 Chr.2: 27082948-27082948 on Build GRCh38
rs201663523 Chr.2: 27082956-27082956 on Build GRCh38
rs200453685 Chr.2: 27082824-27082824 on Build GRCh38
rs754865997 Chr.2: 27082968-27082968 on Build GRCh38
rs753533275 Chr.2: 27082950-27082951 on Build GRCh38
rs745775728 Chr.2: 27082870-27082870 on Build GRCh38
rs755757014 Chr.2: 27082834-27082834 on Build GRCh38
rs376755900 Chr.2: 27082876-27082876 on Build GRCh38
rs905079374 Chr.2: 27082918-27082920 on Build GRCh38

View Full Product Details