Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101774843-101775102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF8
Entrez Gene ID
2253
Gene Name
fibroblast growth factor 8
Gene Aliases
AIGF, FGF-8, HBGF-8, HH6, KAL6
UniGene
Hs.57710
Gene Chromosome Location
Chr.10: 101770130-101780369 on Build GRCh38
Transcripts
NM_001206389.1 NM_006119.4 NM_033163.3 NM_033164.3 NM_033165.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
914059 c.158T>G p.V53G Chr.10: 101774911-101774911 on Build GRCh38
1296860 c.225C>G p.L75L Chr.10: 101774844-101774844 on Build GRCh38
6518212 c.173C>T p.S58L Chr.10: 101774896-101774896 on Build GRCh38
6128181 c.195G>C p.V65V Chr.10: 101774874-101774874 on Build GRCh38
6247376 c.220C>A p.Q74K Chr.10: 101774849-101774849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188846574 Chr.10: 101775007-101775007 on Build GRCh38
rs891119484 Chr.10: 101775027-101775027 on Build GRCh38
rs1048103260 Chr.10: 101774970-101774970 on Build GRCh38
rs940424846 Chr.10: 101775031-101775031 on Build GRCh38
rs753766864 Chr.10: 101774956-101774956 on Build GRCh38
rs767008028 Chr.10: 101774907-101774907 on Build GRCh38
rs929274371 Chr.10: 101774966-101774966 on Build GRCh38
rs754969584 Chr.10: 101774996-101774996 on Build GRCh38
rs754659347 Chr.10: 101774958-101774958 on Build GRCh38
rs778809649 Chr.10: 101775012-101775012 on Build GRCh38
rs909094366 Chr.10: 101774977-101774977 on Build GRCh38
rs538012694 Chr.10: 101775073-101775073 on Build GRCh38
rs990686415 Chr.10: 101774978-101774978 on Build GRCh38
rs769041438 Chr.10: 101774905-101774910 on Build GRCh38
rs752266134 Chr.10: 101774919-101774919 on Build GRCh38
rs923038266 Chr.10: 101775048-101775048 on Build GRCh38
rs765230725 Chr.10: 101774959-101774959 on Build GRCh38
rs535034436 Chr.10: 101774971-101774971 on Build GRCh38
rs1038811169 Chr.10: 101774998-101774998 on Build GRCh38
rs753693921 Chr.10: 101774943-101774943 on Build GRCh38
rs760297996 Chr.10: 101774924-101774924 on Build GRCh38
rs997319184 Chr.10: 101775013-101775013 on Build GRCh38
rs941875635 Chr.10: 101774991-101774991 on Build GRCh38
rs564054337 Chr.10: 101775072-101775072 on Build GRCh38
rs975834701 Chr.10: 101775033-101775033 on Build GRCh38
rs369992360 Chr.10: 101774934-101774934 on Build GRCh38
rs747977350 Chr.10: 101775023-101775023 on Build GRCh38
rs369975105 Chr.10: 101775053-101775053 on Build GRCh38
rs571747403 Chr.10: 101775034-101775034 on Build GRCh38
rs952004334 Chr.10: 101775006-101775006 on Build GRCh38

View Full Product Details