Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26216222-26216495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H2BG
Entrez Gene ID
8339
Gene Name
histone cluster 1, H2bg
Gene Aliases
H2B.1A, H2B/a, H2BFA, dJ221C16.8
UniGene
Hs.591809
Gene Chromosome Location
Chr.6: 26216200-26216644 on Build GRCh38
Transcripts
NM_003518.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7242675 c.374C>A p.S125Y Chr.6: 26216270-26216270 on Build GRCh38
3357991 c.371G>A p.S124N Chr.6: 26216273-26216273 on Build GRCh38
5718369 c.368C>G p.T123R Chr.6: 26216276-26216276 on Build GRCh38
5814875 c.359C>A p.T120N Chr.6: 26216285-26216285 on Build GRCh38
346836 c.282G>A p.E94E Chr.6: 26216362-26216362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779063580 Chr.6: 26216357-26216357 on Build GRCh38
rs746456357 Chr.6: 26216256-26216256 on Build GRCh38
rs770265113 Chr.6: 26216411-26216411 on Build GRCh38
rs770940533 Chr.6: 26216404-26216404 on Build GRCh38
rs367765460 Chr.6: 26216313-26216313 on Build GRCh38
rs41266811 Chr.6: 26216428-26216428 on Build GRCh38
rs748405525 Chr.6: 26216359-26216359 on Build GRCh38
rs772022131 Chr.6: 26216360-26216360 on Build GRCh38
rs1053789424 Chr.6: 26216326-26216326 on Build GRCh38
rs760189973 Chr.6: 26216426-26216426 on Build GRCh38
rs777861065 Chr.6: 26216401-26216401 on Build GRCh38
rs781470850 Chr.6: 26216468-26216468 on Build GRCh38
rs1002799030 Chr.6: 26216322-26216322 on Build GRCh38
rs772404532 Chr.6: 26216398-26216398 on Build GRCh38
rs1034673471 Chr.6: 26216328-26216328 on Build GRCh38
rs765729377 Chr.6: 26216380-26216380 on Build GRCh38
rs752372359 Chr.6: 26216249-26216249 on Build GRCh38
rs371778293 Chr.6: 26216332-26216332 on Build GRCh38
rs761831024 Chr.6: 26216278-26216278 on Build GRCh38
rs372346685 Chr.6: 26216353-26216353 on Build GRCh38
rs1044211051 Chr.6: 26216302-26216302 on Build GRCh38
rs921511542 Chr.6: 26216458-26216458 on Build GRCh38
rs767133701 Chr.6: 26216422-26216422 on Build GRCh38
rs751280874 Chr.6: 26216314-26216314 on Build GRCh38
rs773561876 Chr.6: 26216284-26216284 on Build GRCh38
rs754320179 Chr.6: 26216292-26216292 on Build GRCh38
rs745853955 Chr.6: 26216260-26216260 on Build GRCh38
rs776059675 Chr.6: 26216412-26216412 on Build GRCh38
rs377201962 Chr.6: 26216343-26216343 on Build GRCh38
rs778005479 Chr.6: 26216449-26216449 on Build GRCh38
rs751225846 Chr.6: 26216383-26216383 on Build GRCh38
rs747050536 Chr.6: 26216403-26216403 on Build GRCh38
rs372917727 Chr.6: 26216470-26216470 on Build GRCh38
rs767666925 Chr.6: 26216281-26216281 on Build GRCh38
rs199951228 Chr.6: 26216261-26216261 on Build GRCh38
rs1021191649 Chr.6: 26216352-26216352 on Build GRCh38
rs752469309 Chr.6: 26216437-26216437 on Build GRCh38
rs761370253 Chr.6: 26216416-26216416 on Build GRCh38
rs140919872 Chr.6: 26216473-26216473 on Build GRCh38
rs1048829001 Chr.6: 26216266-26216266 on Build GRCh38
rs138214841 Chr.6: 26216293-26216293 on Build GRCh38
rs548265162 Chr.6: 26216253-26216253 on Build GRCh38
rs759933007 Chr.6: 26216378-26216378 on Build GRCh38
rs967879011 Chr.6: 26216374-26216374 on Build GRCh38
rs768934674 Chr.6: 26216271-26216271 on Build GRCh38
rs761580046 Chr.6: 26216384-26216384 on Build GRCh38
rs139135890 Chr.6: 26216365-26216365 on Build GRCh38
rs748304068 Chr.6: 26216397-26216397 on Build GRCh38
rs772930700 Chr.6: 26216251-26216252 on Build GRCh38
rs774567730 Chr.6: 26216273-26216273 on Build GRCh38
rs780946079 Chr.6: 26216258-26216258 on Build GRCh38
rs570046076 Chr.6: 26216446-26216446 on Build GRCh38
rs375561975 Chr.6: 26216376-26216376 on Build GRCh38
rs777238838 Chr.6: 26216406-26216406 on Build GRCh38
rs772758945 Chr.6: 26216424-26216424 on Build GRCh38
rs778724341 Chr.6: 26216395-26216395 on Build GRCh38
rs201957387 Chr.6: 26216368-26216368 on Build GRCh38
rs949862512 Chr.6: 26216263-26216263 on Build GRCh38
rs143613920 Chr.6: 26216347-26216347 on Build GRCh38
rs765911190 Chr.6: 26216427-26216427 on Build GRCh38
rs766192153 Chr.6: 26216453-26216454 on Build GRCh38
rs887512722 Chr.6: 26216272-26216272 on Build GRCh38
rs779443828 Chr.6: 26216405-26216405 on Build GRCh38
rs368649575 Chr.6: 26216349-26216349 on Build GRCh38
rs755586518 Chr.6: 26216390-26216390 on Build GRCh38
rs763783564 Chr.6: 26216308-26216308 on Build GRCh38
rs958605761 Chr.6: 26216329-26216329 on Build GRCh38
rs996069650 Chr.6: 26216259-26216259 on Build GRCh38
rs186981469 Chr.6: 26216305-26216305 on Build GRCh38
rs200965546 Chr.6: 26216466-26216466 on Build GRCh38
rs374281578 Chr.6: 26216434-26216434 on Build GRCh38
rs750019670 Chr.6: 26216389-26216389 on Build GRCh38
rs369376110 Chr.6: 26216392-26216392 on Build GRCh38
rs772220232 Chr.6: 26216257-26216257 on Build GRCh38
rs766638514 Chr.6: 26216287-26216287 on Build GRCh38
rs777415026 Chr.6: 26216254-26216254 on Build GRCh38
rs747228517 Chr.6: 26216455-26216455 on Build GRCh38
rs775404879 Chr.6: 26216264-26216264 on Build GRCh38
rs756138373 Chr.6: 26216350-26216350 on Build GRCh38
rs16891427 Chr.6: 26216356-26216356 on Build GRCh38
rs887260420 Chr.6: 26216474-26216474 on Build GRCh38
rs746368662 Chr.6: 26216469-26216469 on Build GRCh38
rs914690836 Chr.6: 26216363-26216363 on Build GRCh38
rs202170052 Chr.6: 26216386-26216386 on Build GRCh38
rs760271385 Chr.6: 26216289-26216290 on Build GRCh38
rs973582647 Chr.6: 26216408-26216408 on Build GRCh38
rs182318715 Chr.6: 26216269-26216269 on Build GRCh38
rs762435555 Chr.6: 26216467-26216467 on Build GRCh38

View Full Product Details