Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30986509-30986734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD3B7
Entrez Gene ID
80270
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
Gene Aliases
CBAS1, PFIC4, SDR11E3
UniGene
Hs.460618
Gene Chromosome Location
Chr.16: 30985189-30989152 on Build GRCh38
Transcripts
NM_001142777.1 NM_001142778.1 NM_025193.3 XM_005255601.3 XM_011545960.2 XM_011545961.1 XM_011545962.2 XM_017023732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6323627 c.504G>A p.W168* Chr.16: 30986677-30986677 on Build GRCh38
1324486 c.465C>G p.H155Q Chr.16: 30986638-30986638 on Build GRCh38
5706113 c.433G>A p.G145S Chr.16: 30986606-30986606 on Build GRCh38
3508996 c.421C>T p.P141S Chr.16: 30986521-30986521 on Build GRCh38
5363513 c.438C>T p.N146N Chr.16: 30986611-30986611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903573425 Chr.16: 30986706-30986706 on Build GRCh38
rs765732538 Chr.16: 30986659-30986659 on Build GRCh38
rs104894518 Chr.16: 30986612-30986612 on Build GRCh38
rs560826820 Chr.16: 30986672-30986672 on Build GRCh38
rs370554987 Chr.16: 30986696-30986696 on Build GRCh38
rs768888717 Chr.16: 30986581-30986581 on Build GRCh38
rs779316697 Chr.16: 30986545-30986545 on Build GRCh38
rs762663539 Chr.16: 30986618-30986618 on Build GRCh38
rs773053001 Chr.16: 30986616-30986616 on Build GRCh38
rs748073154 Chr.16: 30986592-30986592 on Build GRCh38
rs751796565 Chr.16: 30986678-30986678 on Build GRCh38
rs539486580 Chr.16: 30986534-30986534 on Build GRCh38
rs912091688 Chr.16: 30986611-30986611 on Build GRCh38
rs150365141 Chr.16: 30986695-30986695 on Build GRCh38
rs780434735 Chr.16: 30986564-30986564 on Build GRCh38
rs766002165 Chr.16: 30986626-30986626 on Build GRCh38
rs774018498 Chr.16: 30986627-30986627 on Build GRCh38
rs776887216 Chr.16: 30986586-30986586 on Build GRCh38
rs750739758 Chr.16: 30986671-30986671 on Build GRCh38
rs749799670 Chr.16: 30986585-30986588 on Build GRCh38
rs151234025 Chr.16: 30986644-30986644 on Build GRCh38
rs894869356 Chr.16: 30986707-30986707 on Build GRCh38
rs780652987 Chr.16: 30986666-30986666 on Build GRCh38
rs757771601 Chr.16: 30986656-30986656 on Build GRCh38
rs758578120 Chr.16: 30986556-30986556 on Build GRCh38
rs747037010 Chr.16: 30986577-30986577 on Build GRCh38
rs777777150 Chr.16: 30986556-30986557 on Build GRCh38
rs771282021 Chr.16: 30986605-30986632 on Build GRCh38
rs755296237 Chr.16: 30986682-30986682 on Build GRCh38
rs759010382 Chr.16: 30986634-30986634 on Build GRCh38
rs927578998 Chr.16: 30986592-30986594 on Build GRCh38
rs766983587 Chr.16: 30986639-30986639 on Build GRCh38
rs750999061 Chr.16: 30986694-30986694 on Build GRCh38
rs746278039 Chr.16: 30986548-30986548 on Build GRCh38
rs140387407 Chr.16: 30986687-30986687 on Build GRCh38

View Full Product Details