Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80107346-80107615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAA
Entrez Gene ID
2548
Gene Name
glucosidase alpha, acid
Gene Aliases
LYAG
UniGene
Hs.1437
Gene Chromosome Location
Chr.17: 80101526-80119882 on Build GRCh38
Transcripts
NM_000152.4 NM_001079803.2 NM_001079804.2 XM_005257193.2 XM_005257194.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1387008 c.728A>G p.D243G Chr.17: 80107592-80107592 on Build GRCh38
3787463 c.711G>A p.A237A Chr.17: 80107575-80107575 on Build GRCh38
5666500 c.729C>A p.D243E Chr.17: 80107593-80107593 on Build GRCh38
1680128 c.701C>T p.T234M Chr.17: 80107565-80107565 on Build GRCh38
3523292 c.735C>T p.F245F Chr.17: 80107599-80107599 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs148578399 Chr.17: 80107590-80107590 on Build GRCh38
rs121907944 Chr.17: 80107574-80107574 on Build GRCh38
rs777964427 Chr.17: 80107570-80107570 on Build GRCh38
rs1015323808 Chr.17: 80107496-80107496 on Build GRCh38
rs539428524 Chr.17: 80107476-80107476 on Build GRCh38
rs74003611 Chr.17: 80107479-80107479 on Build GRCh38
rs752950890 Chr.17: 80107568-80107568 on Build GRCh38
rs763538104 Chr.17: 80107552-80107552 on Build GRCh38
rs79849256 Chr.17: 80107463-80107463 on Build GRCh38
rs745861849 Chr.17: 80107589-80107589 on Build GRCh38
rs771827503 Chr.17: 80107534-80107534 on Build GRCh38
rs773956464 Chr.17: 80107527-80107527 on Build GRCh38
rs760379754 Chr.17: 80107544-80107544 on Build GRCh38
rs200088236 Chr.17: 80107553-80107553 on Build GRCh38
rs528964318 Chr.17: 80107382-80107382 on Build GRCh38
rs977154756 Chr.17: 80107536-80107536 on Build GRCh38
rs976582329 Chr.17: 80107539-80107539 on Build GRCh38
rs766931184 Chr.17: 80107564-80107564 on Build GRCh38
rs2304846 Chr.17: 80107569-80107569 on Build GRCh38
rs1047100216 Chr.17: 80107456-80107456 on Build GRCh38
rs370502229 Chr.17: 80107509-80107509 on Build GRCh38
rs892652135 Chr.17: 80107403-80107403 on Build GRCh38
rs536976019 Chr.17: 80107530-80107530 on Build GRCh38
rs547495337 Chr.17: 80107416-80107416 on Build GRCh38
rs1009726895 Chr.17: 80107404-80107404 on Build GRCh38
rs368328598 Chr.17: 80107575-80107575 on Build GRCh38
rs775090947 Chr.17: 80107542-80107542 on Build GRCh38
rs899274314 Chr.17: 80107394-80107394 on Build GRCh38
rs975301817 Chr.17: 80107484-80107484 on Build GRCh38
rs752054011 Chr.17: 80107565-80107565 on Build GRCh38
rs778882374 Chr.17: 80107579-80107579 on Build GRCh38
rs781731240 Chr.17: 80107567-80107567 on Build GRCh38
rs1005297878 Chr.17: 80107493-80107493 on Build GRCh38
rs561501762 Chr.17: 80107369-80107369 on Build GRCh38
rs952374058 Chr.17: 80107480-80107480 on Build GRCh38
rs78855075 Chr.17: 80107508-80107508 on Build GRCh38
rs1053872423 Chr.17: 80107370-80107370 on Build GRCh38
rs1026293295 Chr.17: 80107412-80107412 on Build GRCh38
rs755337511 Chr.17: 80107566-80107566 on Build GRCh38

View Full Product Details