Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52780634-52780903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPNAT1
Entrez Gene ID
64841
Gene Name
glucosamine-phosphate N-acetyltransferase 1
Gene Aliases
GNA1, GNPNAT, Gpnat1
UniGene
Hs.478025
Gene Chromosome Location
Chr.14: 52775193-52791668 on Build GRCh38
Transcripts
NM_198066.3 XM_005268012.2 XM_006720238.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1477625 c.407+1G>T p.? Chr.14: 52780678-52780678 on Build GRCh38
5478703 c.379G>T p.E127* Chr.14: 52780707-52780707 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760291894 Chr.14: 52780847-52780851 on Build GRCh38
rs572139287 Chr.14: 52780850-52780850 on Build GRCh38
rs945033399 Chr.14: 52780765-52780765 on Build GRCh38
rs766184249 Chr.14: 52780844-52780844 on Build GRCh38
rs767781255 Chr.14: 52780664-52780664 on Build GRCh38
rs781239334 Chr.14: 52780767-52780767 on Build GRCh38
rs779520863 Chr.14: 52780726-52780726 on Build GRCh38
rs746559238 Chr.14: 52780749-52780749 on Build GRCh38
rs775654192 Chr.14: 52780669-52780669 on Build GRCh38
rs750716573 Chr.14: 52780702-52780702 on Build GRCh38
rs369130828 Chr.14: 52780823-52780823 on Build GRCh38
rs903620017 Chr.14: 52780822-52780822 on Build GRCh38
rs374339421 Chr.14: 52780772-52780772 on Build GRCh38
rs917209235 Chr.14: 52780819-52780821 on Build GRCh38
rs765577863 Chr.14: 52780700-52780700 on Build GRCh38
rs764695387 Chr.14: 52780678-52780678 on Build GRCh38
rs766636090 Chr.14: 52780714-52780716 on Build GRCh38
rs757671863 Chr.14: 52780694-52780694 on Build GRCh38
rs760624444 Chr.14: 52780671-52780671 on Build GRCh38
rs111408452 Chr.14: 52780769-52780769 on Build GRCh38
rs751607075 Chr.14: 52780755-52780755 on Build GRCh38
rs371406305 Chr.14: 52780680-52780680 on Build GRCh38
rs754413755 Chr.14: 52780756-52780756 on Build GRCh38
rs868050554 Chr.14: 52780846-52780846 on Build GRCh38
rs112594134 Chr.14: 52780747-52780747 on Build GRCh38
rs758056533 Chr.14: 52780706-52780706 on Build GRCh38
rs758679966 Chr.14: 52780676-52780677 on Build GRCh38

View Full Product Details