Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32948785-32949012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DMA
Entrez Gene ID
3108
Gene Name
major histocompatibility complex, class II, DM alpha
Gene Aliases
D6S222E, DMA, HLADM, RING6
UniGene
Hs.728759
Gene Chromosome Location
Chr.6: 32948614-32953122 on Build GRCh38
Transcripts
NM_006120.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5654754 c.782A>C p.D261A Chr.6: 32948868-32948868 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1020617825 Chr.6: 32948944-32948944 on Build GRCh38
rs771480013 Chr.6: 32948912-32948926 on Build GRCh38
rs550054876 Chr.6: 32948955-32948955 on Build GRCh38
rs1011526382 Chr.6: 32948939-32948939 on Build GRCh38
rs967359482 Chr.6: 32948926-32948928 on Build GRCh38
rs148396755 Chr.6: 32948866-32948866 on Build GRCh38
rs201203849 Chr.6: 32948889-32948889 on Build GRCh38
rs1049703948 Chr.6: 32948864-32948864 on Build GRCh38
rs760838967 Chr.6: 32948856-32948856 on Build GRCh38
rs940360961 Chr.6: 32948839-32948839 on Build GRCh38
rs769591581 Chr.6: 32948826-32948826 on Build GRCh38
rs999507946 Chr.6: 32948952-32948952 on Build GRCh38
rs2395297 Chr.6: 32948920-32948920 on Build GRCh38
rs751972511 Chr.6: 32948884-32948884 on Build GRCh38
rs778387867 Chr.6: 32948882-32948882 on Build GRCh38
rs759762045 Chr.6: 32948868-32948868 on Build GRCh38
rs145946216 Chr.6: 32948923-32948923 on Build GRCh38
rs540515990 Chr.6: 32948959-32948959 on Build GRCh38
rs930842150 Chr.6: 32948810-32948810 on Build GRCh38
rs762732073 Chr.6: 32948833-32948833 on Build GRCh38
rs745477791 Chr.6: 32948818-32948818 on Build GRCh38
rs1032370966 Chr.6: 32948956-32948956 on Build GRCh38
rs129654 Chr.6: 32948922-32948922 on Build GRCh38
rs768697796 Chr.6: 32948834-32948834 on Build GRCh38
rs766137750 Chr.6: 32948912-32948914 on Build GRCh38
rs184563028 Chr.6: 32948984-32948984 on Build GRCh38
rs886730347 Chr.6: 32948811-32948811 on Build GRCh38
rs766321060 Chr.6: 32948858-32948858 on Build GRCh38
rs778723605 Chr.6: 32948832-32948833 on Build GRCh38
rs375343803 Chr.6: 32948830-32948830 on Build GRCh38
rs1001503678 Chr.6: 32948921-32948921 on Build GRCh38
rs745525804 Chr.6: 32948917-32948917 on Build GRCh38
rs201706107 Chr.6: 32948874-32948874 on Build GRCh38
rs549558564 Chr.6: 32948890-32948890 on Build GRCh38
rs367902530 Chr.6: 32948871-32948871 on Build GRCh38
rs538684284 Chr.6: 32948968-32948968 on Build GRCh38

View Full Product Details