Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 85621811-85622064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLRT2
Entrez Gene ID
23768
Gene Name
fibronectin leucine rich transmembrane protein 2
Gene Aliases
-
UniGene
Hs.533710
Gene Chromosome Location
Chr.14: 85530067-85628696 on Build GRCh38
Transcripts
NM_013231.4 XM_005267490.3 XM_011536611.2 XM_017021129.1 XM_017021130.1 XM_017021131.1 XM_017021132.1 XM_017021133.1 XM_017021134.1 XM_017021135.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1252576 c.397T>C p.L133L Chr.14: 85621911-85621911 on Build GRCh38
1207250 c.352G>T p.E118* Chr.14: 85621866-85621866 on Build GRCh38
5482560 c.531G>T p.G177G Chr.14: 85622045-85622045 on Build GRCh38
147827 c.332T>C p.V111A Chr.14: 85621846-85621846 on Build GRCh38
4758408 c.386T>C p.L129P Chr.14: 85621900-85621900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769582808 Chr.14: 85621945-85621945 on Build GRCh38
rs142516343 Chr.14: 85621911-85621911 on Build GRCh38
rs766298483 Chr.14: 85621998-85621998 on Build GRCh38
rs779460062 Chr.14: 85621909-85621909 on Build GRCh38
rs199559498 Chr.14: 85621946-85621946 on Build GRCh38
rs766109044 Chr.14: 85621988-85621988 on Build GRCh38
rs780318340 Chr.14: 85621891-85621891 on Build GRCh38
rs755442700 Chr.14: 85621899-85621899 on Build GRCh38
rs139640372 Chr.14: 85621971-85621971 on Build GRCh38
rs554986569 Chr.14: 85621882-85621882 on Build GRCh38
rs190684718 Chr.14: 85621925-85621925 on Build GRCh38
rs140724546 Chr.14: 85621970-85621970 on Build GRCh38
rs747226438 Chr.14: 85621931-85621931 on Build GRCh38
rs996509714 Chr.14: 85621951-85621951 on Build GRCh38
rs748669304 Chr.14: 85622000-85622000 on Build GRCh38
rs750040176 Chr.14: 85621981-85621981 on Build GRCh38
rs754192129 Chr.14: 85621989-85621989 on Build GRCh38
rs778764956 Chr.14: 85621915-85621915 on Build GRCh38
rs117507441 Chr.14: 85621833-85621833 on Build GRCh38
rs755405673 Chr.14: 85621991-85621991 on Build GRCh38
rs764123585 Chr.14: 85621960-85621960 on Build GRCh38
rs749697862 Chr.14: 85621894-85621894 on Build GRCh38
rs777746741 Chr.14: 85622040-85622040 on Build GRCh38
rs758456805 Chr.14: 85622036-85622036 on Build GRCh38
rs762882941 Chr.14: 85621947-85621947 on Build GRCh38

View Full Product Details