Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17771697-17771940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC1
Entrez Gene ID
3746
Gene Name
potassium voltage-gated channel subfamily C member 1
Gene Aliases
EPM7, KV3.1, KV4, NGK2
UniGene
Hs.552896
Gene Chromosome Location
Chr.11: 17734810-17783055 on Build GRCh38
Transcripts
NM_001112741.1 NM_004976.4 XM_011520080.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4427549 c.745G>A p.E249K Chr.11: 17771839-17771839 on Build GRCh38
2092233 c.806A>G p.N269S Chr.11: 17771900-17771900 on Build GRCh38
925534 c.832C>A p.L278I Chr.11: 17771926-17771926 on Build GRCh38
925533 c.802C>T p.P268S Chr.11: 17771896-17771896 on Build GRCh38
6215818 c.744C>T p.I248I Chr.11: 17771838-17771838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112489382 Chr.11: 17771888-17771888 on Build GRCh38
rs752437957 Chr.11: 17771787-17771787 on Build GRCh38
rs868512387 Chr.11: 17771812-17771812 on Build GRCh38
rs763794003 Chr.11: 17771795-17771795 on Build GRCh38
rs774087624 Chr.11: 17771776-17771776 on Build GRCh38
rs138647448 Chr.11: 17771856-17771856 on Build GRCh38
rs1050543354 Chr.11: 17771746-17771746 on Build GRCh38
rs775885483 Chr.11: 17771790-17771790 on Build GRCh38
rs775163829 Chr.11: 17771882-17771882 on Build GRCh38
rs753431522 Chr.11: 17771910-17771910 on Build GRCh38
rs747207268 Chr.11: 17771748-17771748 on Build GRCh38
rs2229007 Chr.11: 17771838-17771838 on Build GRCh38
rs143670270 Chr.11: 17771811-17771811 on Build GRCh38
rs150993414 Chr.11: 17771803-17771803 on Build GRCh38
rs769612184 Chr.11: 17771760-17771760 on Build GRCh38
rs745968113 Chr.11: 17771730-17771730 on Build GRCh38
rs73424032 Chr.11: 17771771-17771771 on Build GRCh38
rs563524153 Chr.11: 17771809-17771809 on Build GRCh38
rs759363166 Chr.11: 17771868-17771868 on Build GRCh38
rs147271572 Chr.11: 17771844-17771844 on Build GRCh38
rs772094650 Chr.11: 17771878-17771878 on Build GRCh38
rs762606123 Chr.11: 17771767-17771767 on Build GRCh38
rs991751914 Chr.11: 17771829-17771829 on Build GRCh38
rs373113740 Chr.11: 17771810-17771810 on Build GRCh38
rs772042572 Chr.11: 17771734-17771734 on Build GRCh38
rs773018717 Chr.11: 17771766-17771766 on Build GRCh38
rs768887549 Chr.11: 17771835-17771835 on Build GRCh38
rs200015927 Chr.11: 17771804-17771804 on Build GRCh38
rs376576774 Chr.11: 17771896-17771896 on Build GRCh38
rs747047818 Chr.11: 17771831-17771831 on Build GRCh38
rs955213839 Chr.11: 17771749-17771749 on Build GRCh38
rs780073632 Chr.11: 17771817-17771817 on Build GRCh38
rs760208987 Chr.11: 17771892-17771892 on Build GRCh38
rs368573465 Chr.11: 17771900-17771900 on Build GRCh38
rs200873319 Chr.11: 17771777-17771777 on Build GRCh38
rs552970887 Chr.11: 17771735-17771735 on Build GRCh38

View Full Product Details