Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43147835-43148068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM183A
Entrez Gene ID
440585
Gene Name
family with sequence similarity 183 member A
Gene Aliases
-
UniGene
Hs.657614
Gene Chromosome Location
Chr.1: 43142959-43156396 on Build GRCh38
Transcripts
NM_001101376.2 XM_005270875.4 XM_005270876.4 XM_017001322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6498157 c.31C>G p.P11A Chr.1: 43147982-43147982 on Build GRCh38
5430254 c.97_98delCT p.Y34fs*23 Chr.1: 43148048-43148049 on Build GRCh38
4682779 c.71A>G p.Y24C Chr.1: 43148022-43148022 on Build GRCh38
4753936 c.85C>T p.R29* Chr.1: 43148036-43148036 on Build GRCh38
1206217 c.115G>A p.V39M Chr.1: 43148066-43148066 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755756602 Chr.1: 43147924-43147924 on Build GRCh38
rs547550210 Chr.1: 43148012-43148012 on Build GRCh38
rs896848053 Chr.1: 43147933-43147933 on Build GRCh38
rs774181462 Chr.1: 43147998-43147998 on Build GRCh38
rs567779337 Chr.1: 43148037-43148037 on Build GRCh38
rs773192932 Chr.1: 43147976-43147976 on Build GRCh38
rs781265901 Chr.1: 43147958-43147958 on Build GRCh38
rs766708343 Chr.1: 43148039-43148039 on Build GRCh38
rs368259924 Chr.1: 43147954-43147954 on Build GRCh38
rs751979290 Chr.1: 43147936-43147936 on Build GRCh38
rs370647959 Chr.1: 43147915-43147915 on Build GRCh38
rs529636323 Chr.1: 43147876-43147876 on Build GRCh38
rs747499589 Chr.1: 43147948-43147948 on Build GRCh38
rs543048586 Chr.1: 43147893-43147893 on Build GRCh38
rs751124476 Chr.1: 43147908-43147908 on Build GRCh38
rs766399185 Chr.1: 43147907-43147907 on Build GRCh38
rs560912393 Chr.1: 43147871-43147871 on Build GRCh38
rs773900658 Chr.1: 43147976-43147977 on Build GRCh38
rs938492438 Chr.1: 43147872-43147895 on Build GRCh38
rs957546531 Chr.1: 43147947-43147947 on Build GRCh38
rs1051565164 Chr.1: 43147886-43147886 on Build GRCh38
rs779303356 Chr.1: 43147925-43147925 on Build GRCh38
rs775048560 Chr.1: 43148020-43148020 on Build GRCh38
rs371268044 Chr.1: 43148028-43148028 on Build GRCh38
rs753543595 Chr.1: 43148021-43148021 on Build GRCh38
rs759465006 Chr.1: 43148000-43148000 on Build GRCh38
rs201791768 Chr.1: 43147944-43147944 on Build GRCh38
rs767008891 Chr.1: 43147922-43147922 on Build GRCh38
rs759116130 Chr.1: 43147984-43147985 on Build GRCh38
rs199757639 Chr.1: 43147935-43147935 on Build GRCh38
rs541657597 Chr.1: 43147923-43147923 on Build GRCh38
rs981788845 Chr.1: 43147995-43147995 on Build GRCh38
rs770404225 Chr.1: 43147974-43147976 on Build GRCh38
rs770812913 Chr.1: 43147982-43147982 on Build GRCh38
rs763703205 Chr.1: 43148033-43148033 on Build GRCh38
rs141446506 Chr.1: 43147910-43147911 on Build GRCh38
rs200795631 Chr.1: 43147912-43147912 on Build GRCh38
rs748570670 Chr.1: 43147911-43147935 on Build GRCh38
rs770128393 Chr.1: 43147975-43147975 on Build GRCh38
rs989730780 Chr.1: 43147855-43147855 on Build GRCh38
rs888269228 Chr.1: 43147911-43147911 on Build GRCh38
rs374348118 Chr.1: 43148035-43148035 on Build GRCh38
rs565748870 Chr.1: 43147973-43147973 on Build GRCh38
rs373248817 Chr.1: 43147978-43147978 on Build GRCh38
rs34090219 Chr.1: 43147999-43148000 on Build GRCh38
rs890028239 Chr.1: 43147887-43147887 on Build GRCh38
rs1046522679 Chr.1: 43147889-43147889 on Build GRCh38
rs56741253 Chr.1: 43147865-43147865 on Build GRCh38
rs766066148 Chr.1: 43147966-43147966 on Build GRCh38

View Full Product Details