Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110518400-110518645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNA10
Entrez Gene ID
3744
Gene Name
potassium voltage-gated channel subfamily A member 10
Gene Aliases
Kcn1, Kv1.8
UniGene
Hs.248140
Gene Chromosome Location
Chr.1: 110517217-110519175 on Build GRCh38
Transcripts
NM_005549.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6877314 c.147G>A p.G49G Chr.1: 110518641-110518641 on Build GRCh38
5992326 c.343C>T p.R115W Chr.1: 110518445-110518445 on Build GRCh38
5905404 c.353G>A p.R118K Chr.1: 110518435-110518435 on Build GRCh38
5917229 c.256C>T p.Q86* Chr.1: 110518532-110518532 on Build GRCh38
4917200 c.368A>G p.D123G Chr.1: 110518420-110518420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768218340 Chr.1: 110518608-110518608 on Build GRCh38
rs944309494 Chr.1: 110518566-110518566 on Build GRCh38
rs921489394 Chr.1: 110518581-110518581 on Build GRCh38
rs200254580 Chr.1: 110518529-110518529 on Build GRCh38
rs374575317 Chr.1: 110518508-110518508 on Build GRCh38
rs764710770 Chr.1: 110518434-110518434 on Build GRCh38
rs148829307 Chr.1: 110518606-110518606 on Build GRCh38
rs557447296 Chr.1: 110518569-110518569 on Build GRCh38
rs34454042 Chr.1: 110518434-110518434 on Build GRCh38
rs912222512 Chr.1: 110518556-110518556 on Build GRCh38
rs761518775 Chr.1: 110518553-110518553 on Build GRCh38
rs776884567 Chr.1: 110518626-110518626 on Build GRCh38
rs774028773 Chr.1: 110518541-110518541 on Build GRCh38
rs150116384 Chr.1: 110518602-110518602 on Build GRCh38
rs773796325 Chr.1: 110518610-110518610 on Build GRCh38
rs543046084 Chr.1: 110518605-110518605 on Build GRCh38
rs144111598 Chr.1: 110518587-110518587 on Build GRCh38
rs761380272 Chr.1: 110518620-110518620 on Build GRCh38
rs779142461 Chr.1: 110518478-110518478 on Build GRCh38
rs140742991 Chr.1: 110518539-110518539 on Build GRCh38
rs771302509 Chr.1: 110518567-110518567 on Build GRCh38
rs141128096 Chr.1: 110518450-110518450 on Build GRCh38
rs139316514 Chr.1: 110518471-110518471 on Build GRCh38
rs750806540 Chr.1: 110518445-110518445 on Build GRCh38
rs770699258 Chr.1: 110518515-110518515 on Build GRCh38
rs758592054 Chr.1: 110518570-110518570 on Build GRCh38
rs371428121 Chr.1: 110518516-110518516 on Build GRCh38
rs1281174 Chr.1: 110518440-110518440 on Build GRCh38
rs530499570 Chr.1: 110518527-110518527 on Build GRCh38
rs147506944 Chr.1: 110518487-110518487 on Build GRCh38
rs766671278 Chr.1: 110518565-110518565 on Build GRCh38
rs769703694 Chr.1: 110518528-110518528 on Build GRCh38
rs140343405 Chr.1: 110518439-110518439 on Build GRCh38
rs759514030 Chr.1: 110518432-110518432 on Build GRCh38
rs150009335 Chr.1: 110518514-110518514 on Build GRCh38
rs1002743758 Chr.1: 110518494-110518494 on Build GRCh38
rs754364867 Chr.1: 110518455-110518455 on Build GRCh38
rs113755109 Chr.1: 110518502-110518502 on Build GRCh38
rs150926393 Chr.1: 110518444-110518444 on Build GRCh38
rs140376600 Chr.1: 110518572-110518572 on Build GRCh38
rs868838468 Chr.1: 110518588-110518588 on Build GRCh38
rs754168137 Chr.1: 110518567-110518567 on Build GRCh38
rs368764831 Chr.1: 110518523-110518523 on Build GRCh38
rs148190888 Chr.1: 110518448-110518448 on Build GRCh38
rs751787719 Chr.1: 110518576-110518576 on Build GRCh38
rs555576853 Chr.1: 110518509-110518509 on Build GRCh38
rs375380259 Chr.1: 110518625-110518625 on Build GRCh38
rs369996397 Chr.1: 110518507-110518507 on Build GRCh38
rs757442247 Chr.1: 110518585-110518585 on Build GRCh38
rs376192714 Chr.1: 110518524-110518524 on Build GRCh38
rs769742463 Chr.1: 110518603-110518603 on Build GRCh38

View Full Product Details