Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 14608683-14608918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM105A
Entrez Gene ID
54491
Gene Name
family with sequence similarity 105 member A
Gene Aliases
NET20
UniGene
Hs.155085
Gene Chromosome Location
Chr.5: 14581782-14616180 on Build GRCh38
Transcripts
NM_019018.2 XM_011514056.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3612054 c.735C>T p.F245F Chr.5: 14608855-14608855 on Build GRCh38
1063876 c.703C>T p.L235L Chr.5: 14608823-14608823 on Build GRCh38
181436 c.686delT p.S231fs*13 Chr.5: 14608806-14608806 on Build GRCh38
5559072 c.744G>A p.L248L Chr.5: 14608864-14608864 on Build GRCh38
4927431 c.759A>C p.E253D Chr.5: 14608879-14608879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767519708 Chr.5: 14608821-14608821 on Build GRCh38
rs755806851 Chr.5: 14608845-14608845 on Build GRCh38
rs150604883 Chr.5: 14608810-14608810 on Build GRCh38
rs773511077 Chr.5: 14608807-14608807 on Build GRCh38
rs569686210 Chr.5: 14608794-14608794 on Build GRCh38
rs892695664 Chr.5: 14608783-14608783 on Build GRCh38
rs975452624 Chr.5: 14608755-14608755 on Build GRCh38
rs766748110 Chr.5: 14608811-14608811 on Build GRCh38
rs143807003 Chr.5: 14608762-14608762 on Build GRCh38
rs74815459 Chr.5: 14608875-14608875 on Build GRCh38
rs747905234 Chr.5: 14608750-14608750 on Build GRCh38
rs780173763 Chr.5: 14608716-14608716 on Build GRCh38
rs151125975 Chr.5: 14608780-14608780 on Build GRCh38
rs751669584 Chr.5: 14608819-14608819 on Build GRCh38
rs754936440 Chr.5: 14608736-14608736 on Build GRCh38
rs142880301 Chr.5: 14608740-14608740 on Build GRCh38
rs749475791 Chr.5: 14608815-14608815 on Build GRCh38
rs777223750 Chr.5: 14608757-14608757 on Build GRCh38
rs942755777 Chr.5: 14608753-14608753 on Build GRCh38
rs778060841 Chr.5: 14608770-14608770 on Build GRCh38
rs752156539 Chr.5: 14608803-14608803 on Build GRCh38
rs141442803 Chr.5: 14608835-14608835 on Build GRCh38
rs559855564 Chr.5: 14608791-14608791 on Build GRCh38
rs745313527 Chr.5: 14608745-14608745 on Build GRCh38
rs541384204 Chr.5: 14608775-14608775 on Build GRCh38
rs777763123 Chr.5: 14608804-14608804 on Build GRCh38
rs764712957 Chr.5: 14608716-14608718 on Build GRCh38
rs751755840 Chr.5: 14608717-14608717 on Build GRCh38
rs552375238 Chr.5: 14608834-14608834 on Build GRCh38
rs960927010 Chr.5: 14608869-14608869 on Build GRCh38
rs375644202 Chr.5: 14608767-14608767 on Build GRCh38
rs777610369 Chr.5: 14608864-14608864 on Build GRCh38
rs1046537035 Chr.5: 14608816-14608816 on Build GRCh38

View Full Product Details