Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70463596-70463835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUK
Entrez Gene ID
197258
Gene Name
fucokinase
Gene Aliases
1110046B12Rik
UniGene
Hs.7907
Gene Chromosome Location
Chr.16: 70454421-70480274 on Build GRCh38
Transcripts
NM_145059.2 XM_006721162.1 XM_011522928.1 XM_011522929.1 XM_011522930.2 XM_011522931.2 XM_017023012.1 XM_017023013.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6667793 c.122G>A p.G41E Chr.16: 70463662-70463662 on Build GRCh38
6377624 c.146A>G p.D49G Chr.16: 70463686-70463686 on Build GRCh38
5786782 c.164G>T p.G55V Chr.16: 70463704-70463704 on Build GRCh38
6320526 c.96G>T p.R32R Chr.16: 70463636-70463636 on Build GRCh38
1379298 c.125C>T p.T42M Chr.16: 70463665-70463665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569139301 Chr.16: 70463665-70463665 on Build GRCh38
rs533929498 Chr.16: 70463760-70463760 on Build GRCh38
rs888513535 Chr.16: 70463644-70463644 on Build GRCh38
rs200386683 Chr.16: 70463791-70463791 on Build GRCh38
rs754851794 Chr.16: 70463696-70463696 on Build GRCh38
rs925675698 Chr.16: 70463673-70463673 on Build GRCh38
rs369355398 Chr.16: 70463698-70463698 on Build GRCh38
rs746067626 Chr.16: 70463646-70463646 on Build GRCh38
rs780144767 Chr.16: 70463787-70463787 on Build GRCh38
rs766134440 Chr.16: 70463686-70463686 on Build GRCh38
rs150311768 Chr.16: 70463735-70463735 on Build GRCh38
rs774825950 Chr.16: 70463720-70463720 on Build GRCh38
rs373425489 Chr.16: 70463709-70463709 on Build GRCh38
rs749663031 Chr.16: 70463663-70463663 on Build GRCh38
rs757254446 Chr.16: 70463641-70463641 on Build GRCh38
rs776392728 Chr.16: 70463740-70463740 on Build GRCh38
rs866811411 Chr.16: 70463704-70463704 on Build GRCh38
rs747072933 Chr.16: 70463635-70463635 on Build GRCh38
rs977742696 Chr.16: 70463777-70463777 on Build GRCh38
rs748549974 Chr.16: 70463809-70463809 on Build GRCh38
rs1006068228 Chr.16: 70463670-70463670 on Build GRCh38
rs371950142 Chr.16: 70463692-70463692 on Build GRCh38
rs201023038 Chr.16: 70463708-70463708 on Build GRCh38
rs772529735 Chr.16: 70463729-70463729 on Build GRCh38
rs770073301 Chr.16: 70463649-70463649 on Build GRCh38
rs140143924 Chr.16: 70463631-70463631 on Build GRCh38
rs760399324 Chr.16: 70463679-70463679 on Build GRCh38
rs377727682 Chr.16: 70463767-70463767 on Build GRCh38
rs369275877 Chr.16: 70463764-70463764 on Build GRCh38
rs775768249 Chr.16: 70463656-70463656 on Build GRCh38
rs779982094 Chr.16: 70463624-70463624 on Build GRCh38
rs367811053 Chr.16: 70463620-70463620 on Build GRCh38
rs748587910 Chr.16: 70463726-70463726 on Build GRCh38
rs766089906 Chr.16: 70463738-70463738 on Build GRCh38
rs745954428 Chr.16: 70463699-70463699 on Build GRCh38
rs781385329 Chr.16: 70463643-70463643 on Build GRCh38
rs750449972 Chr.16: 70463778-70463778 on Build GRCh38
rs768975149 Chr.16: 70463719-70463719 on Build GRCh38
rs767154340 Chr.16: 70463617-70463617 on Build GRCh38
rs779016102 Chr.16: 70463634-70463634 on Build GRCh38
rs759313314 Chr.16: 70463742-70463742 on Build GRCh38
rs550622605 Chr.16: 70463683-70463683 on Build GRCh38
rs756213310 Chr.16: 70463784-70463784 on Build GRCh38
rs375564886 Chr.16: 70463666-70463666 on Build GRCh38
rs753752470 Chr.16: 70463627-70463627 on Build GRCh38
rs764931038 Chr.16: 70463758-70463758 on Build GRCh38
rs200603725 Chr.16: 70463727-70463727 on Build GRCh38
rs750010776 Chr.16: 70463619-70463619 on Build GRCh38
rs558228101 Chr.16: 70463761-70463761 on Build GRCh38
rs60470914 Chr.16: 70463678-70463678 on Build GRCh38
rs766462388 Chr.16: 70463796-70463796 on Build GRCh38

View Full Product Details