Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49071939-49072204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNA7
Entrez Gene ID
3743
Gene Name
potassium voltage-gated channel subfamily A member 7
Gene Aliases
HAK6, KV1.7
UniGene
Hs.306973
Gene Chromosome Location
Chr.19: 49067418-49072941 on Build GRCh38
Transcripts
NM_031886.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7103822 c.411G>C p.E137D Chr.19: 49072175-49072175 on Build GRCh38
7103821 c.508G>A p.D170N Chr.19: 49072078-49072078 on Build GRCh38
6456100 c.521G>A p.G174D Chr.19: 49072065-49072065 on Build GRCh38
1681200 c.463G>A p.V155I Chr.19: 49072123-49072123 on Build GRCh38
1564317 c.539C>T p.A180V Chr.19: 49072047-49072047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748689298 Chr.19: 49072061-49072061 on Build GRCh38
rs545142998 Chr.19: 49072089-49072089 on Build GRCh38
rs752656469 Chr.19: 49072017-49072018 on Build GRCh38
rs77059600 Chr.19: 49072047-49072047 on Build GRCh38
rs907585876 Chr.19: 49071959-49071959 on Build GRCh38
rs910062669 Chr.19: 49072014-49072014 on Build GRCh38
rs780165257 Chr.19: 49072042-49072042 on Build GRCh38
rs747137703 Chr.19: 49072059-49072059 on Build GRCh38
rs201446279 Chr.19: 49072031-49072031 on Build GRCh38
rs1021402162 Chr.19: 49071967-49071967 on Build GRCh38
rs979894887 Chr.19: 49071975-49071976 on Build GRCh38
rs773978360 Chr.19: 49072091-49072091 on Build GRCh38
rs984387947 Chr.19: 49072087-49072087 on Build GRCh38
rs910093701 Chr.19: 49072127-49072127 on Build GRCh38
rs142677554 Chr.19: 49072139-49072139 on Build GRCh38
rs771444666 Chr.19: 49072172-49072172 on Build GRCh38
rs771427075 Chr.19: 49072178-49072178 on Build GRCh38
rs767405906 Chr.19: 49072051-49072051 on Build GRCh38
rs770221707 Chr.19: 49072046-49072046 on Build GRCh38
rs373588713 Chr.19: 49072124-49072124 on Build GRCh38
rs778668555 Chr.19: 49072175-49072175 on Build GRCh38
rs918883202 Chr.19: 49071993-49071993 on Build GRCh38
rs781342550 Chr.19: 49072148-49072148 on Build GRCh38
rs761183102 Chr.19: 49071999-49071999 on Build GRCh38
rs755070365 Chr.19: 49072060-49072060 on Build GRCh38
rs773687492 Chr.19: 49072083-49072083 on Build GRCh38
rs777648850 Chr.19: 49072063-49072064 on Build GRCh38
rs1014797689 Chr.19: 49072032-49072032 on Build GRCh38
rs534650525 Chr.19: 49072049-49072049 on Build GRCh38
rs115128299 Chr.19: 49071984-49071984 on Build GRCh38
rs940347382 Chr.19: 49071962-49071962 on Build GRCh38
rs758595605 Chr.19: 49072138-49072138 on Build GRCh38
rs376468263 Chr.19: 49072005-49072005 on Build GRCh38
rs775224902 Chr.19: 49072115-49072115 on Build GRCh38
rs767086504 Chr.19: 49072114-49072114 on Build GRCh38
rs1039793635 Chr.19: 49072022-49072022 on Build GRCh38
rs760722550 Chr.19: 49072118-49072118 on Build GRCh38
rs201539045 Chr.19: 49072153-49072153 on Build GRCh38
rs780075778 Chr.19: 49072058-49072058 on Build GRCh38
rs535278472 Chr.19: 49071985-49072005 on Build GRCh38
rs950997183 Chr.19: 49072066-49072066 on Build GRCh38
rs754944257 Chr.19: 49072147-49072147 on Build GRCh38
rs753133537 Chr.19: 49072150-49072150 on Build GRCh38
rs755026437 Chr.19: 49072030-49072030 on Build GRCh38
rs759069301 Chr.19: 49072108-49072108 on Build GRCh38
rs775937500 Chr.19: 49071996-49071996 on Build GRCh38
rs1037343005 Chr.19: 49071971-49071971 on Build GRCh38
rs753673748 Chr.19: 49072126-49072126 on Build GRCh38
rs755825423 Chr.19: 49072052-49072052 on Build GRCh38
rs199663901 Chr.19: 49072157-49072157 on Build GRCh38
rs745521579 Chr.19: 49072177-49072177 on Build GRCh38
rs768993631 Chr.19: 49072001-49072001 on Build GRCh38
rs748946301 Chr.19: 49072088-49072088 on Build GRCh38
rs536830816 Chr.19: 49072037-49072037 on Build GRCh38
rs577674481 Chr.19: 49072055-49072055 on Build GRCh38
rs764843610 Chr.19: 49072009-49072009 on Build GRCh38
rs751489237 Chr.19: 49072028-49072028 on Build GRCh38
rs369215044 Chr.19: 49072019-49072019 on Build GRCh38
rs371711587 Chr.19: 49071990-49071990 on Build GRCh38
rs758231145 Chr.19: 49072035-49072036 on Build GRCh38
rs769999721 Chr.19: 49072062-49072062 on Build GRCh38
rs984315829 Chr.19: 49072011-49072011 on Build GRCh38
rs757480933 Chr.19: 49072036-49072036 on Build GRCh38
rs750629552 Chr.19: 49072057-49072057 on Build GRCh38
rs777972236 Chr.19: 49072155-49072155 on Build GRCh38
rs775130529 Chr.19: 49072184-49072184 on Build GRCh38
rs750107860 Chr.19: 49072016-49072016 on Build GRCh38
rs758414427 Chr.19: 49072040-49072040 on Build GRCh38
rs200579675 Chr.19: 49072043-49072043 on Build GRCh38
rs553009723 Chr.19: 49072050-49072050 on Build GRCh38
rs777028770 Chr.19: 49072044-49072044 on Build GRCh38

View Full Product Details