Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55982105-55982354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBAS
Entrez Gene ID
2631
Gene Name
glioblastoma amplified sequence
Gene Aliases
NIPSNAP2
UniGene
Hs.591069
Gene Chromosome Location
Chr.7: 55964577-56000182 on Build GRCh38
Transcripts
NM_001202469.1 NM_001483.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1158003 c.403C>T p.P135S Chr.7: 55982239-55982239 on Build GRCh38
5031521 c.391G>A p.E131K Chr.7: 55982227-55982227 on Build GRCh38
5662314 c.411C>T p.L137L Chr.7: 55982247-55982247 on Build GRCh38
6044085 c.402T>A p.Y134* Chr.7: 55982238-55982238 on Build GRCh38
4959359 c.444G>A p.K148K Chr.7: 55982280-55982280 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs190182078 Chr.7: 55982256-55982256 on Build GRCh38
rs564003419 Chr.7: 55982164-55982164 on Build GRCh38
rs199850047 Chr.7: 55982237-55982237 on Build GRCh38
rs182076369 Chr.7: 55982177-55982177 on Build GRCh38
rs759145241 Chr.7: 55982311-55982311 on Build GRCh38
rs752142915 Chr.7: 55982313-55982313 on Build GRCh38
rs777773662 Chr.7: 55982229-55982229 on Build GRCh38
rs370376520 Chr.7: 55982321-55982321 on Build GRCh38
rs77829445 Chr.7: 55982261-55982261 on Build GRCh38
rs535466489 Chr.7: 55982238-55982238 on Build GRCh38
rs374505411 Chr.7: 55982289-55982289 on Build GRCh38
rs768976163 Chr.7: 55982267-55982267 on Build GRCh38
rs765682848 Chr.7: 55982290-55982290 on Build GRCh38
rs767175825 Chr.7: 55982312-55982312 on Build GRCh38
rs755954356 Chr.7: 55982226-55982226 on Build GRCh38
rs368639343 Chr.7: 55982204-55982204 on Build GRCh38
rs144366493 Chr.7: 55982225-55982225 on Build GRCh38
rs755633094 Chr.7: 55982251-55982251 on Build GRCh38
rs889986819 Chr.7: 55982286-55982286 on Build GRCh38
rs777259811 Chr.7: 55982281-55982281 on Build GRCh38
rs765942404 Chr.7: 55982175-55982175 on Build GRCh38
rs66485869 Chr.7: 55982297-55982297 on Build GRCh38
rs748414809 Chr.7: 55982185-55982185 on Build GRCh38
rs754457934 Chr.7: 55982188-55982188 on Build GRCh38
rs755718711 Chr.7: 55982318-55982318 on Build GRCh38
rs143364643 Chr.7: 55982250-55982250 on Build GRCh38
rs757643077 Chr.7: 55982165-55982165 on Build GRCh38
rs549792039 Chr.7: 55982199-55982199 on Build GRCh38
rs376656557 Chr.7: 55982240-55982240 on Build GRCh38

View Full Product Details