Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171107618-171107883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO3
Entrez Gene ID
2328
Gene Name
flavin containing monooxygenase 3
Gene Aliases
FMOII, TMAU, dJ127D3.1
UniGene
Hs.445350
Gene Chromosome Location
Chr.1: 171090873-171117819 on Build GRCh38
Transcripts
NM_001002294.2 NM_001319173.1 NM_001319174.1 NM_006894.5 XM_011509345.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
899478 c.322-1G>T p.? Chr.1: 171107674-171107674 on Build GRCh38
5378392 c.410C>T p.S137L Chr.1: 171107763-171107763 on Build GRCh38
5618417 c.472G>A p.E158K Chr.1: 171107825-171107825 on Build GRCh38
3478236 c.374G>A p.W125* Chr.1: 171107727-171107727 on Build GRCh38
3789078 c.367G>A p.G123S Chr.1: 171107720-171107720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572533923 Chr.1: 171107809-171107809 on Build GRCh38
rs1800822 Chr.1: 171107794-171107794 on Build GRCh38
rs749915808 Chr.1: 171107819-171107819 on Build GRCh38
rs745483246 Chr.1: 171107761-171107761 on Build GRCh38
rs1036658969 Chr.1: 171107799-171107799 on Build GRCh38
rs766722862 Chr.1: 171107713-171107713 on Build GRCh38
rs373952198 Chr.1: 171107663-171107663 on Build GRCh38
rs768566952 Chr.1: 171107772-171107772 on Build GRCh38
rs28363546 Chr.1: 171107746-171107746 on Build GRCh38
rs12072582 Chr.1: 171107747-171107747 on Build GRCh38
rs776967590 Chr.1: 171107796-171107796 on Build GRCh38
rs1008342400 Chr.1: 171107725-171107725 on Build GRCh38
rs778868969 Chr.1: 171107838-171107838 on Build GRCh38
rs564555482 Chr.1: 171107831-171107831 on Build GRCh38
rs913978834 Chr.1: 171107683-171107683 on Build GRCh38
rs958294066 Chr.1: 171107681-171107681 on Build GRCh38
rs772225527 Chr.1: 171107784-171107784 on Build GRCh38
rs779808827 Chr.1: 171107765-171107765 on Build GRCh38
rs555554127 Chr.1: 171107697-171107697 on Build GRCh38
rs748979138 Chr.1: 171107835-171107835 on Build GRCh38
rs370477258 Chr.1: 171107779-171107779 on Build GRCh38
rs774093562 Chr.1: 171107775-171107775 on Build GRCh38
rs72549326 Chr.1: 171107811-171107811 on Build GRCh38
rs755417548 Chr.1: 171107735-171107735 on Build GRCh38
rs996915425 Chr.1: 171107672-171107672 on Build GRCh38
rs77738152 Chr.1: 171107769-171107769 on Build GRCh38
rs2266782 Chr.1: 171107825-171107825 on Build GRCh38
rs762847643 Chr.1: 171107668-171107668 on Build GRCh38
rs201646858 Chr.1: 171107740-171107740 on Build GRCh38
rs376881697 Chr.1: 171107682-171107682 on Build GRCh38
rs903835118 Chr.1: 171107652-171107652 on Build GRCh38
rs113006768 Chr.1: 171107720-171107720 on Build GRCh38
rs780703747 Chr.1: 171107759-171107759 on Build GRCh38
rs767713766 Chr.1: 171107704-171107704 on Build GRCh38
rs146587449 Chr.1: 171107764-171107764 on Build GRCh38
rs951676823 Chr.1: 171107781-171107781 on Build GRCh38
rs201673576 Chr.1: 171107767-171107767 on Build GRCh38
rs879572807 Chr.1: 171107746-171107747 on Build GRCh38
rs886045560 Chr.1: 171107783-171107783 on Build GRCh38
rs190217183 Chr.1: 171107766-171107766 on Build GRCh38
rs774300685 Chr.1: 171107711-171107711 on Build GRCh38
rs757188439 Chr.1: 171107684-171107685 on Build GRCh38
rs748055020 Chr.1: 171107842-171107842 on Build GRCh38
rs750611884 Chr.1: 171107706-171107706 on Build GRCh38
rs778836325 Chr.1: 171107773-171107773 on Build GRCh38
rs186763441 Chr.1: 171107694-171107694 on Build GRCh38
rs766237338 Chr.1: 171107801-171107801 on Build GRCh38
rs754675637 Chr.1: 171107837-171107837 on Build GRCh38
rs753154683 Chr.1: 171107802-171107802 on Build GRCh38
rs72549325 Chr.1: 171107795-171107795 on Build GRCh38
rs200164532 Chr.1: 171107737-171107737 on Build GRCh38
rs866756635 Chr.1: 171107742-171107742 on Build GRCh38
rs754119312 Chr.1: 171107719-171107719 on Build GRCh38
rs183161037 Chr.1: 171107800-171107800 on Build GRCh38
rs778624465 Chr.1: 171107825-171107825 on Build GRCh38
rs985370164 Chr.1: 171107647-171107647 on Build GRCh38

View Full Product Details