Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 5045542-5045767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNA5
Entrez Gene ID
3741
Gene Name
potassium voltage-gated channel subfamily A member 5
Gene Aliases
ATFB7, HCK1, HK2, HPCN1, KV1.5, PCN1
UniGene
Hs.150208
Gene Chromosome Location
Chr.12: 5043919-5046788 on Build GRCh38
Transcripts
NM_002234.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4936029 c.1578A>T p.E526D Chr.12: 5045725-5045725 on Build GRCh38
4529402 c.1609G>A p.E537K Chr.12: 5045756-5045756 on Build GRCh38
4919070 c.1477G>A p.G493S Chr.12: 5045624-5045624 on Build GRCh38
6344312 c.1452G>T p.G484G Chr.12: 5045599-5045599 on Build GRCh38
4469085 c.1580C>A p.T527K Chr.12: 5045727-5045727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774013325 Chr.12: 5045683-5045683 on Build GRCh38
rs888485152 Chr.12: 5045566-5045566 on Build GRCh38
rs754755935 Chr.12: 5045653-5045653 on Build GRCh38
rs915470188 Chr.12: 5045735-5045735 on Build GRCh38
rs540903138 Chr.12: 5045744-5045744 on Build GRCh38
rs766020985 Chr.12: 5045596-5045596 on Build GRCh38
rs764615241 Chr.12: 5045632-5045632 on Build GRCh38
rs750495839 Chr.12: 5045725-5045725 on Build GRCh38
rs762575510 Chr.12: 5045587-5045587 on Build GRCh38
rs748807927 Chr.12: 5045668-5045668 on Build GRCh38
rs866813248 Chr.12: 5045610-5045610 on Build GRCh38
rs770511622 Chr.12: 5045669-5045669 on Build GRCh38
rs961406972 Chr.12: 5045663-5045663 on Build GRCh38
rs376656991 Chr.12: 5045720-5045720 on Build GRCh38
rs371569271 Chr.12: 5045619-5045619 on Build GRCh38
rs748151274 Chr.12: 5045656-5045656 on Build GRCh38
rs17221812 Chr.12: 5045742-5045742 on Build GRCh38
rs776978070 Chr.12: 5045693-5045693 on Build GRCh38
rs121908591 Chr.12: 5045727-5045727 on Build GRCh38
rs772634120 Chr.12: 5045666-5045666 on Build GRCh38
rs761415972 Chr.12: 5045560-5045560 on Build GRCh38
rs746053809 Chr.12: 5045588-5045589 on Build GRCh38
rs762348584 Chr.12: 5045695-5045695 on Build GRCh38
rs17221805 Chr.12: 5045644-5045644 on Build GRCh38
rs760979131 Chr.12: 5045630-5045630 on Build GRCh38
rs751776245 Chr.12: 5045734-5045734 on Build GRCh38
rs866495913 Chr.12: 5045597-5045597 on Build GRCh38
rs757749649 Chr.12: 5045633-5045633 on Build GRCh38
rs140711603 Chr.12: 5045654-5045654 on Build GRCh38
rs923695301 Chr.12: 5045745-5045745 on Build GRCh38
rs765368941 Chr.12: 5045635-5045635 on Build GRCh38
rs71582899 Chr.12: 5045743-5045743 on Build GRCh38
rs145832242 Chr.12: 5045686-5045686 on Build GRCh38
rs895545730 Chr.12: 5045746-5045746 on Build GRCh38
rs769367518 Chr.12: 5045572-5045572 on Build GRCh38
rs772762096 Chr.12: 5045573-5045573 on Build GRCh38
rs766516121 Chr.12: 5045728-5045728 on Build GRCh38
rs145163163 Chr.12: 5045640-5045640 on Build GRCh38
rs769845264 Chr.12: 5045659-5045659 on Build GRCh38
rs576837393 Chr.12: 5045641-5045641 on Build GRCh38
rs985758860 Chr.12: 5045723-5045723 on Build GRCh38
rs1029805458 Chr.12: 5045710-5045710 on Build GRCh38

View Full Product Details