Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40743932-40744205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN3
Entrez Gene ID
55177
Gene Name
regulator of microtubule dynamics 3
Gene Aliases
FAM82A2, FAM82C, RMD-3, RMD3, ptpip51
UniGene
Hs.511067
Gene Chromosome Location
Chr.15: 40735884-40755336 on Build GRCh38
Transcripts
NM_001304802.1 NM_001323894.1 NM_001323895.1 NM_001323896.1 NM_001323897.1 NM_018145.2 XM_011521755.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
961294 c.836G>A p.R279H Chr.15: 40744121-40744121 on Build GRCh38
6767036 c.835C>T p.R279C Chr.15: 40744122-40744122 on Build GRCh38
5660823 c.902C>G p.A301G Chr.15: 40744055-40744055 on Build GRCh38
6767033 c.873T>C p.T291T Chr.15: 40744084-40744084 on Build GRCh38
337413 c.843C>T p.A281A Chr.15: 40744114-40744114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960787411 Chr.15: 40744094-40744094 on Build GRCh38
rs752859914 Chr.15: 40744163-40744164 on Build GRCh38
rs757361415 Chr.15: 40744123-40744123 on Build GRCh38
rs772358523 Chr.15: 40744046-40744046 on Build GRCh38
rs367898986 Chr.15: 40744170-40744170 on Build GRCh38
rs747812998 Chr.15: 40744097-40744097 on Build GRCh38
rs762948930 Chr.15: 40744000-40744000 on Build GRCh38
rs769787841 Chr.15: 40744077-40744079 on Build GRCh38
rs770466009 Chr.15: 40744168-40744168 on Build GRCh38
rs756377364 Chr.15: 40744019-40744019 on Build GRCh38
rs767808314 Chr.15: 40744010-40744010 on Build GRCh38
rs777358508 Chr.15: 40744147-40744147 on Build GRCh38
rs772717365 Chr.15: 40744105-40744105 on Build GRCh38
rs753978472 Chr.15: 40744122-40744122 on Build GRCh38
rs762395512 Chr.15: 40744106-40744106 on Build GRCh38
rs779676067 Chr.15: 40744022-40744022 on Build GRCh38
rs386783359 Chr.15: 40743951-40743953 on Build GRCh38
rs374707390 Chr.15: 40744032-40744032 on Build GRCh38
rs879224035 Chr.15: 40743998-40743998 on Build GRCh38
rs780445595 Chr.15: 40744090-40744090 on Build GRCh38
rs376809191 Chr.15: 40744002-40744002 on Build GRCh38
rs199751632 Chr.15: 40744072-40744072 on Build GRCh38
rs752898367 Chr.15: 40744014-40744014 on Build GRCh38
rs761724018 Chr.15: 40744073-40744077 on Build GRCh38
rs368599677 Chr.15: 40744117-40744117 on Build GRCh38
rs188268723 Chr.15: 40744098-40744098 on Build GRCh38
rs201577678 Chr.15: 40744096-40744096 on Build GRCh38
rs899696457 Chr.15: 40744078-40744078 on Build GRCh38
rs371839811 Chr.15: 40744120-40744120 on Build GRCh38
rs753962117 Chr.15: 40744056-40744056 on Build GRCh38
rs759483521 Chr.15: 40744113-40744113 on Build GRCh38
rs900673597 Chr.15: 40743971-40743971 on Build GRCh38
rs769453407 Chr.15: 40744146-40744146 on Build GRCh38
rs900653728 Chr.15: 40744183-40744183 on Build GRCh38
rs775881556 Chr.15: 40744112-40744112 on Build GRCh38
rs778057195 Chr.15: 40744053-40744053 on Build GRCh38
rs760912191 Chr.15: 40744007-40744010 on Build GRCh38
rs750943095 Chr.15: 40744139-40744139 on Build GRCh38
rs763040315 Chr.15: 40744097-40744098 on Build GRCh38
rs878949795 Chr.15: 40743994-40743994 on Build GRCh38
rs760766392 Chr.15: 40744180-40744181 on Build GRCh38
rs201035155 Chr.15: 40744140-40744140 on Build GRCh38
rs772363515 Chr.15: 40744084-40744084 on Build GRCh38
rs746277197 Chr.15: 40744073-40744073 on Build GRCh38
rs751386085 Chr.15: 40744004-40744004 on Build GRCh38
rs371332860 Chr.15: 40744177-40744177 on Build GRCh38
rs200641467 Chr.15: 40744071-40744071 on Build GRCh38
rs746621653 Chr.15: 40744034-40744034 on Build GRCh38
rs768397175 Chr.15: 40744036-40744048 on Build GRCh38
rs775153144 Chr.15: 40744180-40744180 on Build GRCh38
rs776250084 Chr.15: 40744063-40744065 on Build GRCh38
rs542334328 Chr.15: 40744079-40744080 on Build GRCh38
rs149870039 Chr.15: 40743997-40743997 on Build GRCh38
rs1015350380 Chr.15: 40744079-40744079 on Build GRCh38
rs201829056 Chr.15: 40744115-40744115 on Build GRCh38
rs144235442 Chr.15: 40744121-40744121 on Build GRCh38
rs146543934 Chr.15: 40744128-40744128 on Build GRCh38
rs747311347 Chr.15: 40744145-40744145 on Build GRCh38
rs374050616 Chr.15: 40744169-40744169 on Build GRCh38
rs754677686 Chr.15: 40744007-40744007 on Build GRCh38
rs568115523 Chr.15: 40744160-40744160 on Build GRCh38
rs767471384 Chr.15: 40744128-40744128 on Build GRCh38
rs757842241 Chr.15: 40744060-40744060 on Build GRCh38

View Full Product Details